Skip to Content
Merck
All Photos(2)

Key Documents

GF82068790

Gadolinium

wire reel, 100mm, diameter 0.5mm, hard, 99.9%

Synonym(s):

Gadolinium, GD005105

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$5,730.00

$5,730.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$5,730.00

About This Item

Linear Formula:
Gd
CAS Number:
Molecular Weight:
157.25
MDL number:
UNSPSC Code:
12141605
PubChem Substance ID:
NACRES:
NA.23

$5,730.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 820-687-90

resistivity

126 μΩ-cm, 20°C

L × diam.

100 mm × 0.5 mm

bp

3273 °C (lit.)

mp

1313 °C (lit.)

density

7.886 g/mL at 25 °C (lit.)

SMILES string

[Gd]

InChI

1S/Gd

InChI key

UIWYJDYFSGRHKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Water-react 3

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brigit den Adel et al.
Atherosclerosis, 225(2), 274-280 (2012-08-14)
MRI using targeted contrast agents (CA) has emerged as a promising technique to study atherothrombotic disease in vivo. Particularly, the use of targeted Gd and lipid-based nanoparticles has enabled detailed in vivo imaging of various molecular markers of atherosclerotic plaque
Kanae Kawai Miyake et al.
AJR. American journal of roentgenology, 203(2), 272-279 (2014-07-24)
The purpose of this study was to assess the diagnostic value of (18)F-FDG PET/CT and MRI in predicting the clinicopathologic subtypes of breast cancer. The cases of 89 patients with mass-type invasive breast cancer who underwent FDG PET/CT and MRI
Stephanie Nougaret et al.
Radiology, 276(3), 797-808 (2015-05-01)
To investigate magnetic resonance (MR) volumetry of endometrial tumors and its association with deep myometrial invasion, tumor grade, and lymphovascular invasion and to assess the value of apparent diffusion coefficient (ADC) histographic analysis of the whole tumor volume for prediction
Ivan Cokic et al.
Circulation. Cardiovascular imaging, 8(8) (2015-08-12)
Recent canines studies have shown that iron deposition within chronic myocardial infarction (CMI) influences the electric behavior of the heart. To date, the link between the iron deposition and malignant ventricular arrhythmias in humans with CMI is unknown. Patients with
Giovanni Antonini et al.
Pain, 155(8), 1464-1471 (2014-05-03)
Although classical trigeminal neuralgia (CTN) is frequently caused by neurovascular contact (NVC) at the trigeminal root entry zone (REZ), both anatomical and MRI studies have shown that NVC of the trigeminal nerve frequently occurs in individuals without CTN. To assess

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service