Merck
  • Phase separation in coamorphous systems: in silico prediction and the experimental challenge of detection.
Phase separation in coamorphous systems: in silico prediction and the experimental challenge of detection.
Molecular pharmaceutics (2014-05-16)
Katja Pajula, Lieke Wittoek, Vesa-Pekka Lehto, Jarkko Ketolainen, Ossi Korhonen
ABSTRACT
Combinatorial chemistry has enabled the production of very potent drugs that might otherwise suffer from poor solubility and low oral bioavailability. One approach to increase solubility is to make the drug amorphous, which leads to problems associated with drug stability. To improve stability, one option is to molecularly disperse the drug in a matrix. However, the primary reason for the failed stabilization with this approach is phase separation, which has been carefully studied in polymeric systems. Nevertheless, the amorphous-amorphous phase separation in coamorphous small molecule mixtures has not yet been reported. The goal of the present study was to experimentally detect the amorphous-amorphous phase separation between two small molecules. A modified in silico method for predicting miscibility by the Flory-Huggins interaction parameter is presented, where conformational variations of the studied molecules were taken into account. A series of drug-drug mixtures, with different mixture ratios, were analyzed by conventional differential scanning calorimetry (DSC(conv)) to detect possible amorphous-amorphous phase separations. The phase separation of coamorphous drug-drug mixtures was also monitored by temperature modulated DSC (MDSC) and Fourier transform infrared (FT-IR) imaging at temperatures above Tg for prolonged time periods. Amorphous-amorphous phase separation was not detected with DSC(conv), probably due to the slow kinetics of phase separation. However, the melting of the separated and subsequently crystallized phases was detected by MDSC. Furthermore, FT-IR imaging was able to detect the separation of the two amorphous phases, which demonstrates the ability of this method to detect small molecule phase separations.
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard