Merck
  • HDAC 1/4-mediated silencing of microRNA-200b promotes chemoresistance in human lung adenocarcinoma cells.
HDAC 1/4-mediated silencing of microRNA-200b promotes chemoresistance in human lung adenocarcinoma cells.
Oncotarget (2014-05-17)
Dong-Qin Chen, Ban-Zhou Pan, Jia-Yuan Huang, Kai Zhang, Shi-Yun Cui, Wei De, Rui Wang, Long-Bang Chen
ABSTRACT
Chemoresistance is one of the most significant obstacles in lung adenocarcinoma (LAD) treatment, and this process involves genetic and epigenetic dysregulation of chemoresistance-related genes. Previously, we have shown that restoration of microRNA (miR)-200b significantly reverses chemoresistance of human LAD cells by targeting E2F3. However, the molecular mechanisms involved in the silencing of miR-200b are still unclear. Here we showed that histone deacetylase (HDAC) inhibitors could restore the expression of miR-200b and reverse chemoresistant phenotypes of docetaxel-resistant LAD cells. HDAC1/4 repression significantly increased miR-200b expression by upregulating histone-H3 acetylation level at the two miR-200b promoters partially via a Sp1-dependent pathway. Furthermore, silencing of HDAC1/4 suppressed cell proliferation, promoted cell apoptosis, induced G2/M cell cycle arrest and ultimately reversed in vitro and in vivo chemoresistance of docetaxel-resistant LAD cells, at least partially in a miR-200b-dependent manner. HDAC1/4 suppression-induced rescue of miR-200b contributed to downregulation of E2F3, survivin and Aurora-A, and upregulation of cleaved-caspase-3. HDAC1/4 levels in docetaxel-insensitive human LAD tissues, inversely correlated with miR-200b, were upregulated compared with docetaxel-sensitive tissues. Taken together, our findings suggest that the HDAC1/4/Sp1/miR-200b/E2F3 pathway is responsible for chemoresistance of docetaxel-resistant LAD cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human HDAC9 (3)
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hdac9
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sp1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Chn1
Sigma-Aldrich
MISSION® esiRNA, targeting human SP1