Skip to Content
Merck
All Photos(1)

Key Documents

EMU173171

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Chn1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$534.00
50 μG
$953.00

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
$534.00
50 μG
$953.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$534.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGAAGATGTCAAGATGGCTTTTGATAGAGATGGTGAGAAGGCGGATATTTCTGTGAACATGTATGAGGACATCAACATTATCACTGGTGCACTTAAACTGTACTTCAGGGATCTGCCAATTCCTCTCATCACATACGATGCCTACCCCAAGTTCATTGAGTCTGCCAAAATTATGGACCCTGACGAGCAATTGGAGACCCTTCACGAAGCACTGAGATCGCTGCCGCCTGCCCACTGCGAGACGCTCCGGTACCTCATGGCGCATCTCAAGAGAGTGACCCTTCATGAGAAGGAGAATCTGATGAGTGCAGAGAACCTTGGGATCGTGTTTGGACCAACCCTCATGAGATCCCCAGAGCTCG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Zeng et al.
American journal of physiology. Endocrinology and metabolism, 307(4), E384-E397 (2014-07-10)
Activation of conventional PKCs (cPKC) is a key signaling that directs the cardiac toxicity of hyperglycemia. AKAP150, a scaffold protein of the A-kinase anchoring proteins (AKAPs) family, is less defined regarding its capability to anchor and regulate cardiac cPKC signaling.
Junlan Feng et al.
Journal of cellular and molecular medicine, 18(10), 2125-2134 (2014-09-19)
Our previously published study documented a deregulation of the microRNA miR-150 in colorectal cancer. Here, we investigated further, in vitro and in vivo, the potential molecular mechanisms underlying the involvement of miR-150 in colorectal cancer, using the appropriate molecular biological
Peng Yang et al.
Cellular signalling, 27(7), 1525-1532 (2015-03-18)
Surgery-induced inflammation has been associated with cancer recurrence and metastasis in colorectal cancer (CRC). As a constituent of gram-negative bacteria, lipopolysaccharide (LPS) is frequently abundant in the peri-operative window. However, the definite roles of LPS in tumour progression remain elusive.
Deguang Xing et al.
Oncology reports, 31(6), 2692-2700 (2014-04-24)
In the present study, we designed and conducted a series of assays to determine the expression of voltage-gated sodium channel (VGSC) neonatal isoform Nav1.5 (nNav1.5) in human brain astrocytoma and its effect on the proliferation, migration, invasion and apoptosis of
Minfei Jin et al.
PloS one, 9(8), e103965-e103965 (2014-08-05)
MicroRNA (miR)-150 has been reported to be dramatically downregulated in human epithelial ovarian cancer (EOC) tissues and patients' serum compared to normal controls. This study aimed to investigate clinical significance and molecular mechanisms of miR-150 in EOC. In the current

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service