Skip to Content
Merck
All Photos(1)

Key Documents

EHU090071

Sigma-Aldrich

MISSION® esiRNA

targeting human HDAC9 (3)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGTTCTCCAGGCTCTGGTCCCAGTTCACCAAACAATGGGCCAACTGGAAGTGTTACTGAAAATGAGACTTCGGTTTTGCCCCCTACCCCTCATGCCGAGCAAATGGTTTCACAGCAACGCATTCTAATTCATGAAGATTCCATGAACCTGCTAAGTCTTTATACCTCTCCTTCTTTGCCCAACATTACCTTGGGGCTTCCCGCAGTGCCATCCCAGCTCAATGCTTCGAATTCACTCAAAGAAAAGCAGAAGTGTGAGACGCAGACGCTTAGGCAAGGTGTTCCTCTGCCTGGGCAGTATGGAGGCAGCATCCCGGCATCTTCCAGCCACCCTCATGTTACTTTAGAGGGAAAGCCACCCAACAGCAGCCACCAGGCTCTCCTGCAGCATTTATTATTGAAAGAACAAATGCGACAGCAAAAGCTT

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaoyan Wang et al.
Molecular carcinogenesis, 59(12), 1392-1408 (2020-10-21)
Countless evidence suggests that long noncoding RNAs (lncRNAs) are involved in human malignant cancers, including esophageal squamous cell carcinoma (ESCC), although their exact function remains unclear. In the present study, we aimed to investigate the roles and molecular mechanisms of the
Zhiqiang Yu et al.
Oncology letters, 17(3), 3296-3304 (2019-03-15)
Gastric cancer (GC) is a common life-threatening cancer type worldwide, with an increasing prevalence and a high rate of mortality. Due to limitations in clinical treatment, surgery has become the most efficient strategy for the treatment of GC. It is
Yue Yang et al.
The Journal of nutritional biochemistry, 40, 172-177 (2016-12-05)
Activation of hepatic stellate cells (HSCs) is critical for liver fibrosis development. Previously, we showed that astaxanthin (ASTX), a xanthophyll carotenoid, has antifibrogenic effects in LX-2 cells, a human HSC cell line. We sought to determine the effect of ASTX
Zhongxing Liang et al.
Cancer chemotherapy and pharmacology, 85(2), 413-423 (2020-01-08)
Although histone deacetylase (HDAC) inhibitors have been shown to effectively induce the inhibition of proliferation and migration in breast cancer, the mechanism of HDAC9's contribution to chemoresistance remains poorly understood. The aim of this study was to investigate the role
Yuxin Li et al.
Cell death & disease, 11(9), 753-753 (2020-09-17)
HDAC inhibitors are efficacious for treating lymphoma, but display limited efficacy in treating solid tumors. Here, we investigated the relationship between HDAC inhibitor resistance and the tumor immune environment in colorectal cancer. Our data indicated that among the investigated immune

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service