Merck
  • Phosphoprotein analysis reveals MEK inhibition as a way to target non-small cell lung cancer tumor initiating cells.
Phosphoprotein analysis reveals MEK inhibition as a way to target non-small cell lung cancer tumor initiating cells.
International journal of radiation biology (2014-03-22)
Lovisa Lundholm, Petra Hååg, Therese Juntti, Rolf Lewensohn, Kristina Viktorsson
ABSTRACT
We aimed to analyze the activation status of commonly deregulated receptor tyrosine kinases (RTK) in human non-small cell lung cancer (NSCLC) tumor initiating cells (TIC) previously demonstrated to be refractory to ionizing radiation and chemotherapy. Phosphorylated RTK and important signaling nodes were assayed using PathScan RTK Signaling Antibody Array Kit in NSCLC TIC and bulk cells 4 h post-irradiation (IR) and validated by Western blot. The effect of mitogen- activated protein kinase kinase (MEK) inhibition combined with IR was analyzed using clonogenic assay. H125 TIC displayed decreased basal phosphorylation of insulin-like growth factor 1 receptor (IGF-1R) and signal transducer and activator of transcription 1 (STAT1) (Tyr701) as compared to bulk cells. Total IGF-1R levels were significantly lower in NSCLC TIC as compared to bulk cells. A higher degree of extracellular signal-regulated kinase (ERK) phosphorylation was evident in TIC and concordantly MEK inhibition reduced TIC viability. Moreover, MEK inhibition also decreased clonogenicity upon IR suggesting that MEK and downstream signaling impart on TIC radiation response. We demonstrate reduced basal phosphorylation of several signaling pathways including lower total IGF-1R levels in NSCLC TIC which is of potential concern for RTK inhibitor use. Importantly, MEK inhibition decreased cell viability of NSCLC TIC alone or in combination with IR.
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
MOPS, BioUltra, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard