Merck
  • Development of a UHPLC-MS/MS method for the determination of plasma histamine in various mammalian species.
Development of a UHPLC-MS/MS method for the determination of plasma histamine in various mammalian species.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-09-30)
Jia Liu, Lei Wang, Wenjuan Hu, Xiaoyan Chen, Dafang Zhong
ABSTRACT
Histamine is an important mediator of anaphylactic reactions. Although several methods have been developed to measure histamine levels, each has its limitations. In this study, we developed and validated a convenient bioanalytical method for the qualitative and quantitative determination of histamine in plasma samples from humans, beagle dogs, Sprague-Dawley rats, and imprinting control region mice. A simple plasma protein precipitation method using acetonitrile was selected, and hydrophilic interaction liquid chromatography coupled with mass spectrometry was used for sample separation and detection. Histamine was subjected to gradient elution with acetonitrile, ammonium acetate buffer, and formic acid. A mass spectrometer equipped with an electrospray ionization source was operated in the positive-ion multiple reaction monitoring mode for the detection of histamine and the internal standard. The [M+H](+) transitions were m/z 112→95 for histamine and m/z 116→99 for d4-histamine, which was used as the internal standard. The lower limit of quantification was 0.2μg/L and the calibration range was 0.2-500μg/L. The overall recovery ranged from 93.6% to 102.8%. The intra- and inter-run precision and accuracy were <15% for plasma samples from all four species. The method was validated by measuring the plasma histamine concentrations in five healthy human volunteers. In conclusion, we have developed and validated a novel bioanalytical method for the quantification of histamine levels in plasma samples from various mammalian species.
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Pyrazole, 98%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Histamine bisphosphate monohydrate
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Histamine bisphosphate monohydrate, ≥99.0% (NT)