Merck
  • Identification of potential pancreatic cancer serum markers: Increased sialyl-Lewis X on ceruloplasmin.
Identification of potential pancreatic cancer serum markers: Increased sialyl-Lewis X on ceruloplasmin.
Clinica chimica acta; international journal of clinical chemistry (2015-01-18)
Meritxell Balmaña, Ariadna Sarrats, Esther Llop, Sílvia Barrabés, Radka Saldova, María José Ferri, Joan Figueras, Esther Fort, Rafael de Llorens, Pauline M Rudd, Rosa Peracaula
ABSTRACT
Pancreatic adenocarcinoma (PDAC) usually shows an enhanced expression of sialyl-Lewis X (sLe(x)) and related epitopes. PDAC may secrete some of the proteins carrying such increased sLe(x) determinant into serum, so they could be used as PDAC markers. Previously, we identified acute-phase proteins with increased sLe(x) in both PDAC and in chronic pancreatitis patients. In this study, depleted sera from the main acute-phase proteins has been analysed for the search of proteins with increased sLe(x) levels in PDAC. Sera from healthy controls, chronic pancreatitis and PDAC patients were depleted, electrophoresed and subjected to sLe(x) immunodetection. Proteins that differentially expressed sLe(x) in PDAC were trypsin digested and identified by LC-ESI-QTOF mass spectrometry. Five protein bands that differentially expressed sLe(x) in PDAC were identified and corresponded to seven different acute-phase proteins. Among them, ceruloplasmin (CP) was selected for further analysis. N-glycan sequencing of CP confirmed the increase of sLe(x) levels in CP in PDAC patients. Healthy controls, chronic pancreatitis and PDAC patients' sera were immunoprecipitated with anti-CP antibodies, and their sLe(x) and CP levels were analysed by western blot. The sLe(x)/CP ratio tended to be higher for the PDAC group, which altogether suggests that the sLe(x)/CP ratio could be a useful biomarker for PDAC.
Sigma-Aldrich
Sodium chloride solution, 0.85%
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Anthranilamide, ≥98%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Supelco
Anthranilamide, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous