Merck
  • Synthesis, molecular structure, biological properties and molecular docking studies on Mn(II), Co(II) and Zn(II) complexes containing bipyridine-azide ligands.
Synthesis, molecular structure, biological properties and molecular docking studies on Mn(II), Co(II) and Zn(II) complexes containing bipyridine-azide ligands.
European journal of medicinal chemistry (2014-12-03)
Vijayan Thamilarasan, Arumugam Jayamani, Nallathambi Sengottuvelan
ABSTRACT
Metal complexes of the type Mn(bpy)2(N3)2 (1), Co(bpy)2(N3)2·3H2O (2) and Zn2(bpy)2(N3)4 (3) (Where bpy = 2,2-bipyridine) have been synthesized and characterized by elemental analysis and spectral (FT-IR, UV-vis) studies. The structure of complexes (1-3) have been determined by single crystal X-ray diffraction studies and the configuration of ligand-coordinated metal(II) ion was well described as distorted octahedral coordination geometry for Mn(II), Co(II) and distorted square pyramidal geometry for Zn(II) complexes. DNA binding interaction of these complexes (1-3) were investigated by UV-vis absorption, fluorescence circular dichroism spectral and molecular docking studies. The intrinsic binding constants Kb of complexes 1, 2 and 3 with CT-DNA obtained from UV-vis absorption studies were 8.37 × 10(4), 2.23 × 10(5) and 5.52 × 10(4) M(-1) respectively. The results indicated that the three complexes are able to bind to DNA with different binding affinity, in the order 2 > 1 > 3. Complexes (1-3) exhibit a good binding propensity to bovine serum albumin (BSA) proteins having relatively high binding constant values. Gel electrophoresis assay demonstrated the ability of the complexes 1-3 promote the cleavage ability of the pBR322 plasmid DNA in the presence of the reducing agent 3-mercaptopropionic acid (MPA) but with different cleavage mechanisms: the complex 3 cleaves DNA via hydrolytic pathway (T4 DNA ligase assay), while the DNA cleavage by complexes 1 and 2 follows oxidative pathway. The chemical nuclease activity follows the order: 2 > 1 > 3. The effects of various activators were also investigated and the nuclease activity efficacy followed the order MPA > GSH > H2O2 > Asc. The cytotoxicity studies of complexes 1-3 were tested in vitro on breast cancer cell line (MCF-7) and they found to be active.
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, 10 mg/mL in H2O
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ethidium bromide solution, ~1% in H2O
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, 500 μg/mL in H2O
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Cobalt(II) acetate tetrahydrate, reagent grade
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Cobalt(II) acetate tetrahydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ethidium bromide, BioReagent, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)