Skip to Content
Merck
All Photos(2)

Key Documents

403024

Sigma-Aldrich

Cobalt(II) acetate tetrahydrate

ACS reagent, ≥98.0%

Synonym(s):

Cobaltous acetate tetrahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$216.00

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$216.00

About This Item

Linear Formula:
(CH3COO)2Co · 4H2O
CAS Number:
Molecular Weight:
249.08
Beilstein:
3731397
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
crystalline powder
granular

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

ACS reagent

Assay

≥98.0%
98.0-102.0% (ACS specification)

form

crystalline powder
granular

reaction suitability

reagent type: catalyst
core: cobalt

impurities

≤0.01% insolubles

anion traces

chloride (Cl-): ≤0.002%
nitrate (NO3-): ≤0.01%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Cu: ≤0.002%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
Na: ≤0.05%
Ni: ≤0.1%
Pb: ≤0.001%
Zn: ≤0.01%

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC(=O)O[Co]OC(C)=O

InChI

1S/2C2H4O2.Co.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

ZBYYWKJVSFHYJL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Cobalt(II) acetate tetrahydrate may be used in the synthesis of the following:
  • cobalt nanoparticles[1]
  • cobalt(II)–aminophenyltetrazolate coordination polymer[2]
  • tricobalt complexes with mixed μ-acetato and μ-pyrazolato ligands[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cobalt nanoparticles synthesis from Co(CH3COO)2 by thermal decomposition.
Shao H, et al.
Journal of magnetism and magnetic materials, 304(1), 28-30 (2006)
Mechanism of thermal decomposition of cobalt acetate tetrahydrate.
Wanjun T and Donghua C.
Chemical Papers, 61(4), 329-332 (2007)
A synthetic strategy for a new series of oxo-centered tricobalt complexes with mixed bridging ligands of acetate and pyrazolate anions.
Yoshida J, et al.
Dalton Transactions, 42(7), 2406-2413 (2013)
Eagleson M
Concise Encyclopedia Chemistry, 241-241 (1994)
Tiffany M Smith et al.
Dalton transactions (Cambridge, England : 2003), 43(19), 7263-7268 (2014-04-01)
The hydrothermal reaction of cobalt(II) acetate tetrahydrate, 4-aminophenyl tetrazole and water at 135 °C for 48 hours provided orange crystals of [Co3(OH)2(H2O)2(aptet)4] (1) (aptet = 4-aminophenyltetrazolate). The three-dimensional structure of compound 1 is constructed from chains of corner- and edge-sharing

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service