Merck
  • Evaluation and comparison of the kinetic performance of ultra-high performance liquid chromatography and high-performance liquid chromatography columns in hydrophilic interaction and reversed-phase liquid chromatography conditions.
Evaluation and comparison of the kinetic performance of ultra-high performance liquid chromatography and high-performance liquid chromatography columns in hydrophilic interaction and reversed-phase liquid chromatography conditions.
Journal of chromatography. A (2014-12-03)
Huiying Song, Erwin Adams, Gert Desmet, Deirdre Cabooter
ABSTRACT
An intrinsic performance comparison is made of the reduction in analysis time that can be obtained when switching from HPLC to UHPLC column formats in HILIC and reversed-phase conditions. A detailed overview of the packing characteristics of both stationary phase types is given first. It is demonstrated that HILIC columns demonstrate higher external porosity values than their reversed-phase counterparts resulting in lower flow resistance values. Column total porosity values determined from the elution time of a small marker molecule are shown to depend strongly on the composition of the mobile phase. To omit errors that might arise from an over- or underestimation of the column void time, all plate height and kinetic plot data are therefore expressed as a function of the interstitial velocity. Although only a limited number of columns are evaluated in this study, it is shown that the column efficiency of the HILIC columns is lower than that of their reversed-phase counterparts, at least for the compounds evaluated here. Despite this lower efficiency, the kinetic performance of both stationary phase types is similar, due to the much lower viscosity of the mobile phases typically used in HILIC conditions. Finally, it is demonstrated that a similar, yet slightly larger reduction in analysis time can be obtained when switching from HPLC column formats to UHPLC formats in HILIC compared to reversed-phase conditions.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thymine, suitable for cell culture, BioReagent
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymine, ≥99%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Cytosine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%