Merck
  • Non-covalent photo-patterning of gelatin matrices using caged collagen mimetic peptides.
Non-covalent photo-patterning of gelatin matrices using caged collagen mimetic peptides.
Macromolecular bioscience (2014-12-06)
Yang Li, Boi Hoa San, Julian L Kessler, Jin Hwan Kim, Qingguo Xu, Justin Hanes, Seungju Michael Yu
ABSTRACT
To address the downside of conventional photo-patterning which can alter the chemical composition of protein scaffolds, we developed a non-covalent photo-patterning strategy for gelatin (denatured collagen) hydrogels that utilizes UV activated triple helical hybridization of caged collagen mimetic peptide (caged CMP). Here we present 2D and 3D photo-patterning of gelatin hydrogels enabled by the caged CMP derivatives, as well as creation of concentration gradients of CMPs. CMP's specificity for binding to gelatin allows patterning of almost any synthetic or natural gelatin-containing matrix, such as gelatin-methacrylate hydrogels and corneal tissues. This is a radically new tool for immobilizing drugs to natural tissues and for functionalizing scaffolds for complex tissue formation.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethanesulfonic acid, 95%
Supelco
Fmoc chloride, derivatization grade (HPLC), LiChropur, ≥99.0% (HPLC)
Sigma-Aldrich
2-Pyrrolidinone, purum, ≥98.0% (GC)
Sigma-Aldrich
Fmoc chloride, BioReagent, ≥99.0% (HPLC)
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
5(6)-Carboxyfluorescein, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Fmoc-Cys(Trt)-OH, ≥95.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
2-Pyrrolidinone, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Fmoc chloride, 97%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Fmoc-Pro-OH, ≥90% (HPLC)
Sigma-Aldrich
2-Pyrrolidinone, 99%
Sigma-Aldrich
5(6)-Carboxyfluorescein, Dye content 90 %
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
2-Nitrobenzaldehyde, 98%
Supelco
1,2-Dichloroethane, analytical standard
Sigma-Aldrich
Piperidine solution, suitable for peptide synthesis, 20% in DMF
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications