Skip to Content
Merck
All Photos(1)

Key Documents

21877

Sigma-Aldrich

5(6)-Carboxyfluorescein

suitable for fluorescence, BioReagent, ≥95% (HPLC)

Synonym(s):

5(6)-FAM, 5,6-CF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H12O7
CAS Number:
Molecular Weight:
376.32
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥95% (HPLC)

form

powder

mp

275 °C (dec.) (lit.)

fluorescence

λex 492 nm; λem 517 nm in 0.1 M Tris pH 8.0

suitability

suitable for fluorescence

SMILES string

OC(=O)c1ccc2c(c1)C(=O)OC23c4ccc(O)cc4Oc5cc(O)ccc35.OC(=O)c6ccc7C(=O)OC8(c9ccc(O)cc9Oc%10cc(O)ccc8%10)c7c6

InChI

1S/2C21H12O7/c22-11-2-5-15-17(8-11)27-18-9-12(23)3-6-16(18)21(15)14-4-1-10(19(24)25)7-13(14)20(26)28-21;22-11-2-5-14-17(8-11)27-18-9-12(23)3-6-15(18)21(14)16-7-10(19(24)25)1-4-13(16)20(26)28-21/h2*1-9,22-23H,(H,24,25)

InChI key

BPVHBBXCESDRKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5(6)-Carboxyfluorescein, also known as 5(6)-FAM, is a fluorescent tracer and available as a mixture of 5-Carboxyfluorescein and 6-Carboxyfluorescein isomers. The fluorescence of 5(6)-Carboxyfluorescein is determined by the xanthene moiety. 5(6)-Carboxyfluorescein has two characteristic properties:
  • It has 2 wavelengths for maximum absorbance (465 and 490nm).
  • And its fluorescence emission, at 515nm(max.), increases as a function of pH in the physiological range of 6-7.4.

Application

5(6)-Carboxyfluorescein is used to characterize tumor cells and normal cells in vivo. It labels proteins derived through N-hydroxysuccinimide (NHS) ester-mediated derivatization. 5(6)-Carboxyfluorescein is suitable for detecting cell viability while developing an in vitro anthelmintic drug assay.

Other Notes

A probe of intracellular pH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mordon et al.
Journal of photochemistry and photobiology. B, Biology, 13(3-4), 307-314 (1992-05-15)
The pH of the interstitial fluid of malignant tumours tends to be lower than that of normal tissue and is depressed by glucose administration. This study aimed to evaluate the effectiveness of dual-wavelength fluorometry using a pH-dependent indicator (5,6-carboxyfluorescein: 5,6-CF)
Charlemagne Gnoula et al.
Talanta, 71(5), 1886-1892 (2007-03-30)
A new in vitro assay for anthelmintic activity using Caenorhabditis elegans is based on the ability of 5(6)-carboxyfluorescein diacetate (CFDA) to indicate the worm's viability. It is shown for the first time that the treatment of a suspension of worms
Jagpreet S Nanda et al.
Methods in enzymology, 536, 87-94 (2014-01-16)
N-hydroxysuccinimde (NHS) ester-mediated derivitization involves the reaction of this amine-reactive group with the primary amines of a protein or a biomolecule. Using NHS chemistry allows one to conjugate various fluorescent probes, biotin, and cross-linkers to primary amines. For example, we
M L Graber et al.
Analytical biochemistry, 156(1), 202-212 (1986-07-01)
We evaluated four different fluoroprobes to determine their capabilities and limitations in measuring intracellular pH by the fluorescent indicator technique. In vitro, carboxyfluorescein, dimethylcarboxyfluorescein, biscarboxyethyl carboxyfluorescein, and 4-methylumbelliferone (4MU) all showed comparably intense fluorescence and excellent pH sensitivity near their
Jorien Claes et al.
Blood, 124(10), 1669-1676 (2014-06-22)
Adhesion of Staphylococcus aureus to blood vessels under shear stress requires von Willebrand factor (VWF). Several bacterial factors have been proposed to interact with VWF, including VWF-binding protein (vWbp), a secreted coagulase that activates the host's prothrombin to generate fibrin.

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service