Skip to Content
Merck
All Photos(3)

Key Documents

447293

Sigma-Aldrich

5(6)-Carboxyfluorescein

Dye content 90 %

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$220.00
5 G
$719.00

$220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$220.00
5 G
$719.00

About This Item

Empirical Formula (Hill Notation):
C21H12O7
CAS Number:
Molecular Weight:
376.32
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥89.5% (HPLC)

Quality Level

form

powder

composition

Dye content, 90%

mp

275 °C (dec.) (lit.)

λmax

492 nm

ε (extinction coefficient)

12134 at 284 nm in 0.1 M NaOH at 0.005 g/L
43060 at 239 nm in 0.1 M NaOH at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OC(=O)c1ccc2c(c1)C(=O)OC23c4ccc(O)cc4Oc5cc(O)ccc35.OC(=O)c6ccc7C(=O)OC8(c9ccc(O)cc9Oc%10cc(O)ccc8%10)c7c6

InChI

1S/2C21H12O7/c22-11-2-5-15-17(8-11)27-18-9-12(23)3-6-16(18)21(15)14-4-1-10(19(24)25)7-13(14)20(26)28-21;22-11-2-5-14-17(8-11)27-18-9-12(23)3-6-15(18)21(14)16-7-10(19(24)25)1-4-13(16)20(26)28-21/h2*1-9,22-23H,(H,24,25)

InChI key

BPVHBBXCESDRKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Foerg et al.
The AAPS journal, 10(2), 349-359 (2008-07-01)
We investigated the metabolic stability of four cell penetrating peptides (CPPs), namely SAP, hCT(9-32)-br, [Palpha] and [Pbeta], when in contact with either subconfluent HeLa, confluent MDCK or Calu-3 epithelial cell cultures. Additionally, through analysis of their cellular translocation efficiency, we
Yang Li et al.
Bioconjugate chemistry, 24(1), 9-16 (2012-12-21)
Although fibrous collagens are major structural components of extracellular matrix in mammals, collagen overproduction is associated with many human diseases including cancers and fibrosis. Collagen is typically identified in biomedical research by Western blot and immunohistochemistry; however, anticollagen antibodies employed
Gabriel Ohlsson et al.
Lab on a chip, 12(22), 4635-4643 (2012-08-17)
Screening assays designed to probe ligand and drug-candidate regulation of membrane proteins responsible for ion-translocation across the cell membrane are wide spread, while efficient means to screen membrane-protein facilitated transport of uncharged solutes are sparse. We report on a microfluidic-based
Jorien Claes et al.
Blood, 124(10), 1669-1676 (2014-06-22)
Adhesion of Staphylococcus aureus to blood vessels under shear stress requires von Willebrand factor (VWF). Several bacterial factors have been proposed to interact with VWF, including VWF-binding protein (vWbp), a secreted coagulase that activates the host's prothrombin to generate fibrin.
Jie Zhou et al.
Talanta, 99, 625-630 (2012-09-13)
A hairpin molecular beacon tagged with carboxyfluorescein in combination with graphene oxide as a quencher reagent was used to detect the DNA damage by chemical reagents. The fluorescence of molecular beacon was quenched sharply by graphene oxide; while in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service