Merck
  • HspB8 is neuroprotective during oxygen glucose deprivation and reperfusion.
HspB8 is neuroprotective during oxygen glucose deprivation and reperfusion.
Current neurovascular research (2015-01-06)
Binbin Yang, He Zhang, Xiaoye Mo, Han Xiao, Zhiping Hu
ABSTRACT
Heat shock protein B8 (HspB8) is a chaperone protein that is highly and constitutively expressed in the brain, cardiac tissue and many other organs. Recently, it has been shown that HspB8 can enhance cardiac function and render cardioprotection. However, the potential benefits of HspB8 action on ischemic stroke and the underlying mechanism(s) are largely unknown. To investigate whether HspB8 exerts protective effects on in vitro ischemia/ reperfusion (I/R) injury, mouse neuroblastoma cells were subjected to oxygen-glucose deprivation and reoxygenation (OGD/R). At first, we investigated and described the HspB8 expression and distribution in N2A cells exposed to OGD/R. Expressions of HspB8 were upregulated in mouse N2A cells after OGD/R, both at mRNA and protein levels. The level of HspB8 began increased after the OGD/R and peaked at 12 hour (12h) after the reperfusion, then declined at 24-hour time points, however, the level of HspB8 was still significantly increased compared to controls. Immunofluorescence analysis revealed that the expressed HspB8 was constitutively localized in the cytoplasm and at the periphery of the nucleus. Nuclear HspB8 levels increased after OGD/R compared with levels in the control, beginning as early as 4h reperfusion, the most conspicuous nuclear HspB8 staining was observed after 24h of reperfusion. Furthermore, overexpression of HspB8 reduced OGD/R -induced apoptosis by reducing the release of cytochrome c from mitochondria to cytosol. In conclusion, our data demonstrated that increased HspB8 expression and its nuclear shift in mouse N2A neuroblastoma cells protected against I/R injury, resulting in reduced apoptosis with the decrease of the release of cytochrome c from mitochondria to cytosol. Therefore, HspB8 might play a fundamental role in opposing and alleviating I/R injury in primary myocardial cells, and it may constitute a new therapeutic target for cerebral ischemic diseases.
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in tissue culture media, DMSO solution
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)