Merck
  • miR-125a-5p is a prognostic biomarker that targets HDAC4 to suppress breast tumorigenesis.
miR-125a-5p is a prognostic biomarker that targets HDAC4 to suppress breast tumorigenesis.
Oncotarget (2014-12-17)
Tsung-Hua Hsieh, Chia-Yi Hsu, Cheng-Fang Tsai, Cheng-Yu Long, Chee-Yin Chai, Ming-Feng Hou, Jau-Nan Lee, Deng-Chyang Wu, Shao-Chun Wang, Eing-Mei Tsai
ABSTRACT
Identifying stably expressed tumor markers that can be used easily to detect cancer is currently an important area of cancer research. By using miRNA microarray, we identified 20 differentially expressed miRNAs in serum samples of breast cancer patients. Expression of miR-125a-5p was relatively lower in patients with shorter survival compared to long-term survivors. In a cohort of breast cancer patients (N = 300), serum expression of miR-125a-5p was negatively and significantly correlated with tumor grade (P = 0.004), lymph-node status (P = 0.004), and tumor size (P < 0.001). Low miR-125a-5p expression was an independent prognostic marker (OR = 0.421; 95% CI = 0.184 to 0.961; P = 0.04) associated with poor survival rates (P = 0.0062). We show that miR-125a-5p directly inhibits expression of the HDAC4 gene, resulting in tumor suppression in vitro and in vivo. Together these results demonstrate that serum miR-125a-5p level in breast cancer may be a useful prognostic biomarker and offer a novel therapeutic avenue by targeting HDAC4 in breast cancer.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard