Merck
  • Regulated stability of eukaryotic elongation factor 2 kinase requires intrinsic but not ongoing activity.
Regulated stability of eukaryotic elongation factor 2 kinase requires intrinsic but not ongoing activity.
The Biochemical journal (2015-02-12)
Xuemin Wang, Jianling Xie, Sergio Regufe da Mota, Claire E Moore, Christopher G Proud
ABSTRACT
Eukaryotic elongation factor 2 kinase (eEF2K) is an atypical protein kinase which negatively regulates protein synthesis, is activated under stress conditions and plays a role in cytoprotection, e.g. in cancer cells. It is regarded as a possible target for therapeutic intervention in solid tumours. Earlier studies showed that eEF2K is degraded by a proteasome-dependent pathway in response to genotoxic stress and that this requires a phosphodegron that includes an autophosphorylation site. Thus, application of eEF2K inhibitors would stabilize eEF2K, partially negating the effects of inhibiting its activity. In the present study, we show that under a range of other stress conditions, including acidosis or treatment of cells with 2-deoxyglucose, eEF2K is also degraded. However, in these settings, the previously identified phosphodegron is not required for its degradation. Nevertheless, kinase-dead and other activity-deficient mutants of eEF2K are stabilized, as is a mutant lacking a critical autophosphorylation site (Thr348 in eEF2K), which is thought to be required for eEF2K and other α-kinases to achieve their active conformations. In contrast, application of small-molecule eEF2K inhibitors does not stabilize the protein. Our data suggest that achieving an active conformation, rather than eEF2K activity per se, is required for its susceptibility to degradation. Additional degrons and E3 ligases beyond those already identified are probably involved in regulating eEF2K levels. Our findings have significant implications for therapeutic targeting of eEF2K, e.g. in oncology.
Product Description
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)