Merck
  • Detection of clenbuterol hydrochloride residuals in pork liver using a customized surface plasmon resonance bioanalyzer.
Detection of clenbuterol hydrochloride residuals in pork liver using a customized surface plasmon resonance bioanalyzer.
PloS one (2015-03-24)
Jiandong Hu, Ruipeng Chen, Shun Wang, Tingting Wang, Yuanyuan Zhao, Jianwei Li, Xinran Hu, Hao Liang, Juanhua Zhu, Xiaohui Sun, Liuzheng Ma, Min Jiang
ABSTRACT
A surface plasmon resonance (SPR) immunoassay with an immobilization of self-assembled molecular identification membrane for the detection of residual Clenbuterol Hydrochloride (CLB) in pork liver was systematically investigated and experimentally validated for its high performance. SPR immunoassay with a regular competitive inhibition assay cannot be directly verified to detect CLB residuals. In this study, the binding of Au film with mercaptopropionic acid was investigated using the known form of the strong S-Au covalent bonds formed by the chemical radical of the mercaptopropionic acid and the Au film. After that, the immunoglobulin IgG of swine (SwIgG-CLB) was bonded with the mercaptopropionic acid by covalent -CO-NH- amide bonding. The modified comprehensive analysis of how the membrane structure works was introduced together with the customized SPR bioanalyzer. In order to evaluate the performance of this biomembrane structure, the concentrations of CLB-contained solutions of 0 ng · mL(-1), 10 ng · mL(-1), 20 ng · mL(-1), 33.3 ng · mL(-1), and 40 ng · mL(-1) were prepared by adding CLB reagents into the solutions of CLB antibody (Clenbuterol Hydrochloride Antibody, CLB-Ab), successively and then the response unit (RU) was measured individually. Using the data collected from the linear CCD array, the fitting curve was established with the R-Square value of 0.9929. Correspondingly, the recovery rate ranged from 88.48% to 103.21% was experimented and the limit of detection of CLB in 1.26 ng · mL(-1) was obtained efficiently. It was concluded that the detection method associated with biomembrane properties is expected to contribute much to the determination of residual CLB in pork liver quantitatively by using the customized SPR bioanalyzer.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
1-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%