Merck
  • Coated dextrin microcapsules of amlodipine incorporable into orally disintegrating tablets for geriatric patients.
Coated dextrin microcapsules of amlodipine incorporable into orally disintegrating tablets for geriatric patients.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2014-12-03)
Dong-Jin Jang, Soo Kyung Bae, Euichaul Oh
ABSTRACT
To improve oral absorption and patient compliance when using amlodipine, novel coated dextrin microcapsules incorporable into orally disintegrating tablets (ODT's) were investigated. Amlodipine-loaded dextrin microcapsules (ADM) were prepared by spray-drying a mixture of amlodipine free base dissolved in ethanol and aqueous dextrin solution. The ADM were suspended in Eudragit(®) EPO solution in ethanol and subsequently spray-dried to collect coated ADM (CADM). The ADM or CADM were blended with ODT excipients and then directly compressed into ODTs. The ADM and CADM used were both spherical with smooth surfaces and had mean particle sizes of 13.3 and 18.5μm, respectively. Amlodipine was dispersed in an amorphous state and was readily encapsulated within ADM or CADM. Unlike the ADM, the tableted CADM remained intact without rupture during tableting, which was consistent with no loss of ethanol (0.82%) entrapped in the ODTs containing the CADM (ODTs-CADM). The amlodipine content appeared to be uniformly maintained as designed in all the dextrin microcapsules and ODTs. The ODTs-CADM compressed with 3kp of hardness showed acceptable ODT characteristics: fast disintegration time (29.8s) and low friability (0.1%). Drug dissolution from the ODTs-CADM was much faster than that of amlodipine free base itself at both pH 1.2 and 6.8 over the tested time. CADM demonstrated significantly higher plasma concentrations (2.7 fold in AUC0-24h and 2.5 fold in Cmax) in SD rats than did amlodipine free base. These results indicate that CADM substantially increased the oral absorption of amlodipine and can be incorporated into ODTs while maintaining their original physicochemical features. The dextrin microcapsules coated using Eudragit(®) EPO may be applied to the development of an amlodipine ODT formulation for improving geriatric patient compliance.
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O