Merck
  • Engineering of nano-crystalline drug suspensions: employing a physico-chemistry based stabilizer selection methodology or approach.
Engineering of nano-crystalline drug suspensions: employing a physico-chemistry based stabilizer selection methodology or approach.
International journal of pharmaceutics (2014-10-04)
Mostafa Nakach, Jean-René Authelin, Tharwat Tadros, Laurence Galet, Alain Chamayou
ABSTRACT
This paper describes a systematic approach to select optimum stabilizer for the preparation of nano-crystalline suspensions of an active pharmaceutical ingredient (API). The stabilizer can be either a dispersant or a combination of dispersant and wetting agent. The proposed screening method is a quick and efficient way to investigate a large number of stabilizers based on the principles of physical-chemistry and employs a stepwise approach. The methodology has been divided in two main parts; the first part being focused on the qualitative screening with the objective of selecting the best candidate(s) for further investigation, the second part has been focused on quantitative screening with the objective to optimize the ratio and amount of wetting and dispersing agents, based on wettability, surface charges measurement, adsorption evaluation, process-ability evaluation and storage stability. The results showed clearly that SDS/PVP 40/60% (w/w) (sodium dodecyl sulfate/poly(vinyl pyrrolidone)) at a total concentration of 1.2% was the optimum stabilizer composition, at which the resulting nanosuspensions were stable for more than 50 days at room temperature.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
6-O-Palmitoyl-L-ascorbic acid, BioXtra, ≥99.0% (RT)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
(±)-Propylene oxide, analytical standard
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Ethylene oxide solution, certified reference material, 50 mg/mL in methanol
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Ascorbic acid 6-palmitate, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Ethylene oxide solution, 2.5-3.3 M in THF
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Ascorbyl Palmitate, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbyl palmitate, European Pharmacopoeia (EP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard