Merck
  • Allelic imbalance at an 8q24 oncogenic SNP is involved in activating MYC in human colorectal cancer.
Allelic imbalance at an 8q24 oncogenic SNP is involved in activating MYC in human colorectal cancer.
Annals of surgical oncology (2014-01-07)
Keishi Sugimachi, Atsushi Niida, Ken Yamamoto, Teppei Shimamura, Seiya Imoto, Hisae Iinuma, Yoshiaki Shinden, Hidetoshi Eguchi, Tomoya Sudo, Masahiko Watanabe, Junichi Tanaka, Shinei Kudo, Kazuo Hase, Masato Kusunoki, Kazutaka Yamada, Yasuhiro Shimada, Kenichi Sugihara, Yoshihiko Maehara, Satoru Miyano, Masaki Mori, Koshi Mimori
ABSTRACT
The rs6983267 at 8q24.21 has been established as a significant cancer-related single nucleotide polymorphism (SNP). The risk allele showed similarity to the binding site of transcription factor TCF4/LEF1 that activates transcription of MYC. However, little is known about the role of this SNP in increasing MYC activity in colorectal cancers (CRCs). The genotypes of rs6983267 in peripheral blood and primary cancers, MYC activity and copy number (CN) alteration were examined in 107 CRCs. Next, we plotted the number of cancers cell lines exhibiting specific G/T genotypes in 746 cancer cell lines of the Sanger Institute database. Then we validated the relationship between the 8q24 SNP status and clinicopathologic parameters in 68 CRCs with loss of heterozygosity (LOH). The MYC module activity was activated by either transcription in the risk allele (G) or by amplification in the non-risk allele (T). Then, we confirmed that the CN amplification dominantly occurred in the non-risk allele, whereas CN neutral LOH, which indicated uniparental disomy (UPD) was more frequently observed for the risk allele. Finally, we confirmed that risk allele dominant cases, either by amplification or by UPD, indicated a more malignant clinical phenotype than non-risk allele dominant cases. The development of CRC requires MYC activation through retention of the risk allele, or amplification of the non-risk allele at the oncogenic SNP in the site of primary tumor.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O