Merck
  • Monitoring of PAEMs and beta-agonists in urine for a small group of experimental subjects and PAEs and beta-agonists in drinking water consumed by the same subjects.
Monitoring of PAEMs and beta-agonists in urine for a small group of experimental subjects and PAEs and beta-agonists in drinking water consumed by the same subjects.
Journal of hazardous materials (2014-03-19)
Saou-Hsing Liou, Gordon C C Yang, Chih-Lung Wang, Yu-Han Chiu
ABSTRACT
This 5-month study contains two parts: (1) to monitor the concentrations of 11 phthalate esters metabolites (PAEMs) and two beta-agonists in human urine samples collected from a small group of consented participants including 16 females and five males; and (2) to analyze the residues of phthalate esters (PAEs) and beta-agonists in various categories of drinking water consumed by the same group of subjects. Each category of human urine and drinking water had 183 samples of its own. The analytical results showed that nine PAEMs were detected in human urine and eight PAEs were detected in drinking water samples. It was found that average concentrations of PAEMs increased as the age increased, but no significant difference between sexes. Further, using the principal component analysis, the loadings of age effect were found to be two times greater than that of gender effect in terms of four DEHP metabolites. Regarding beta-agonists of concern (i.e., ractopamine and salbutamol), they were neither detected in human urine nor drinking water samples in this study.
Product Description
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzyl butyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Ammonium acetate solution, 7.5 M
Sigma-Aldrich
Ammonium acetate, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Diisononyl phthalate, ester content ≥99 % (mixture of C9 isomers), technical grade
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Benzyl butyl phthalate, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Benzyl butyl phthalate, analytical standard
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis