Merck
  • Importance of CRF receptor-mediated mechanisms of the bed nucleus of the stria terminalis in the processing of anxiety and pain.
Importance of CRF receptor-mediated mechanisms of the bed nucleus of the stria terminalis in the processing of anxiety and pain.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2014-05-24)
Lee Tran, Jay Schulkin, Beverley Greenwood-Van Meerveld
ABSTRACT
Corticotropin-releasing factor (CRF)-mediated mechanisms in the bed nucleus of the stria terminalis (BNST) have a pivotal role in stress-induced anxiety and hyperalgesia. Although CRF is known to activate two receptor subtypes, CRF1 and CRF2, attempts to delineate the specific role of each subtype in modulating anxiety and nociception have been inconsistent. Here we test the hypothesis that CRF1 and CRF2 receptor activation in the anteriolateral BNST (BNSTAL) facilitates divergent mechanisms modulating comorbid anxiety and hyperalgesia. Microinfusions of the specific antagonists CP376395 and Astressin2B into the BNSTAL were used to investigate CRF1 and CRF2 receptor functions, respectively. We found that CRF1 and CRF2 receptors in the BNSTAL had opposing effects on exploratory behavior in the elevated plus-maze, somatic mechanical threshold, and the autonomic and endocrine response to stress. However, CRF1 or CRF2 receptor antagonism in the BNSTAL revealed complementary roles in facilitating the acoustic startle and visceromotor reflexes. Our results suggest that the net effect of CRF1 and CRF2 receptor activation in the BNSTAL is pathway-dependent and provides important insight into the CRF receptor-associated circuitry that likely underpins stress-induced pathologies.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard