Merck
  • Synthesis of poly(meth)acrylates with thioether and tertiary sulfonium groups by ARGET ATRP and their use as siRNA delivery agents.
Synthesis of poly(meth)acrylates with thioether and tertiary sulfonium groups by ARGET ATRP and their use as siRNA delivery agents.
Biomacromolecules (2014-12-18)
Matthew C Mackenzie, Arun R Shrivats, Dominik Konkolewicz, Saadyah E Averick, Michael C McDermott, Jeffrey O Hollinger, Krzysztof Matyjaszewski
ABSTRACT
The field of RNA interference depends on the development of safe and efficient carriers for short interfering ribonucleic acid (siRNA) delivery. Conventional cationic monomers for siRNA delivery have utilized the nitrogen heteroatom to produce cationic charges. Here, we polymerized cationic sulfonium (meth)acrylate by activators regenerated by electron transfer (ARGET) atom transfer radical polymerization (ATRP) to form polymers with narrow molecular weight distributions for siRNA delivery. The tertiary sulfonium species was stable toward dealkylation in water but less stable in the polar aprotic solvent dimethyl sulfoxide. Block copolymers poly(ethylene oxide) with poly(meth)acrylate containing sulfonium moieties were prepared as an siRNA delivery platform. Results suggested block copolymers were biocompatible up to 50 μg/mL in vitro and formed polyplexes with siRNA. Additionally, block copolymers protected siRNAs against endonuclease digestion and facilitated knockdown of glyceraldehyde 3-phosphate dehydrogenase (Gapdh) mRNA expression in murine calvarial preosteoblasts. The versatility, biocompatibility, and cationic nature of these tertiary sulfonium groups are expected to find widespread biological applications.
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
2-(Methylthio)ethanol, ≥99%
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Methyl trifluoromethanesulfonate, ≥98%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-(Methylthio)ethanol, 99%
Supelco
Aluminum oxide, for the determination of hydrocarbons
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, basic
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%