Skip to Content
Merck
All Photos(1)

Key Documents

634875

Sigma-Aldrich

Aluminum oxide

single crystal substrate, <0001>

Synonym(s):

Sapphire, Alumina

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$192.00
5 EA
$618.00

$192.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$192.00
5 EA
$618.00

About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
NA.23

$192.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

Crystallographic D spacing ((0001) - c plane: 2.165 Angstrom)
Crystallographic D spacing ((1011) - s plane: 1.961 Angstrom)
Crystallographic D spacing ((1040) - m plane: 1.375 Angstrom)
Crystallographic D spacing ((1102) - r plane: 1.740 Angstrom)
Crystallographic D spacing ((1120) - a plane: 2.379 Angstrom)
Crystallographic D spacing ((1123) - n plane: 1.147 Angstrom)
single side polished

Quality Level

form

single crystal substrate

hardness

9 mohs

L × W × thickness

10 mm × 10 mm × 0.5 mm

mp

2040 °C (lit.)

semiconductor properties

<0001>

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

Common substrate for III-V nitrides as well as many other epitaxial films.

Physical properties

thermal expansion = 7.5 × 10-6 /°C; specific heat = 0.10 cal/°C; thermal conductivity = 46.06 @ 0 °C, 25.12 @ 100 °C, 12.56 @ 400 °C (W/m,K); dielectric constant = ~9.4 @ 300 K at A axis, ~11.58 @ C axis; loss tangent at 10 GHz < 2 × 10-5 at A axis, 5 × 10-6 at C axis

Physical form

Crystalline, hexagonal (a = 4.758 Å, c = 12.992)

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Siddiqi et al.
Dalton transactions (Cambridge, England : 2003), 44(32), 14349-14353 (2015-07-23)
A simple one-step and gram-scale synthesis of [Au5Mes5] from AuCl3 was developed, and this molecular precursor was used to generate Au nanoparticles on SiO2 and Al2O3. While [Au5Mes5] does not react with surface silanols and is only physisorbed, its incipient
Joohoon Kang et al.
ACS nano, 9(4), 3596-3604 (2015-03-19)
Solution dispersions of two-dimensional (2D) black phosphorus (BP)--often referred to as phosphorene--are achieved by solvent exfoliation. These pristine, electronic-grade BP dispersions are produced with anhydrous organic solvents in a sealed-tip ultrasonication system, which circumvents BP degradation that would otherwise occur
Chien-Chih Lin et al.
Nanoscale, 5(17), 8090-8097 (2013-07-25)
We demonstrated a promising route for enhancing temperature sensitivity, improving saturation voltage, and reducing power consumption of the MOS(p) tunneling temperature sensors by introducing ultrathin Al2O3 into the dielectric stacks. Detailed illustrations of the working mechanism and device concept are
Jaakko Akola et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(25), 10129-10134 (2013-06-01)
Glass formation in the CaO-Al2O3 system represents an important phenomenon because it does not contain typical network-forming cations. We have produced structural models of CaO-Al2O3 glasses using combined density functional theory-reverse Monte Carlo simulations and obtained structures that reproduce experiments
Colin J Ingham et al.
Biotechnology advances, 30(5), 1089-1099 (2011-08-23)
Porous aluminum oxide (PAO) is a ceramic formed by an anodization process of pure aluminum that enables the controllable assembly of exceptionally dense and regular nanopores in a planar membrane. As a consequence, PAO has a high porosity, nanopores with

Articles

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service