Skip to Content
Merck
All Photos(8)

Documents

746452

Sigma-Aldrich

Magnesium sulfate

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

Agency

suitable for SM 5210

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

ReagentPlus®
Redi-Dri

Assay

≥99.5%

form

powder

quality

free-flowing

pH

~7.9 (25 °C, 50 g/L)

SMILES string

[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Magnesium sulfate is a magnesium salt with tocolytic ability. Its role to induce an analgesic effect during intraoperative and postoperative periods has been investigated. It is reported to prevent eclampsia. Its ability to remove water from water-miscible solvents by salting-out phase separation has been studied.

Application

Magnesium sulfate may be used as a drying agent for organic extracts.

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lex W Doyle
Current opinion in pediatrics, 24(2), 154-159 (2012-01-10)
Antenatal magnesium sulfate may reduce the excessive rates of cerebral palsy in survivors of very preterm birth. There are five randomized controlled trials of magnesium sulfate therapy given to the mother prior to very preterm birth which have reported neurological
Lex W Doyle et al.
Obstetrics and gynecology, 113(6), 1327-1333 (2009-05-23)
To systematically review rates of neurologic outcomes reported in childhood for the preterm fetus exposed to antenatal magnesium sulfate. We searched the Cochrane Pregnancy and Childbirth Group's Trials Register, CENTRAL (The Cochrane Library 2008, Issue 3), relevant references from retrieved
Robert J Floor et al.
Chembiochem : a European journal of chemical biology, 15(11), 1660-1672 (2014-07-01)
We explored the use of a computational design framework for the stabilization of the haloalkane dehalogenase LinB. Energy calculations, disulfide bond design, molecular dynamics simulations, and rational inspection of mutant structures predicted many stabilizing mutations. Screening of these in small
Claire L O'Brien et al.
Gut, 63(10), 1596-1606 (2014-01-17)
Our aim was to determine whether or not specific microorganisms were transported selectively to lymph nodes in Crohn's disease (CD) by comparing node and mucosal microbial communities in patients and controls. We also sought evidence of dysbiosis and bacterial translocation.
Federica Belluti et al.
ChemMedChem, 9(2), 371-382 (2014-01-10)
A significant improvement in the treatment of trypanosomiases has been achieved with the recent development of nifurtimox-eflornithine combination therapy (NECT). As an alternative to drug combinations and as a means to overcome most of the antitrypanosomatid drug discovery challenges, a

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service