Merck
  • Phosphodiesterase 8a supports HIV-1 replication in macrophages at the level of reverse transcription.
Phosphodiesterase 8a supports HIV-1 replication in macrophages at the level of reverse transcription.
PloS one (2014-10-09)
Thijs Booiman, Viviana Cobos Jiménez, Karel A van Dort, Angélique B van 't Wout, Neeltje A Kootstra
ABSTRACT
HIV-1 infected macrophages play a key role in HIV-1 infection. Even during anti-retroviral treatment, macrophages keep producing virus due to suboptimal tissue penetration and reduced efficacy of antiretrovirals. It is therefore of major importance to understand which host factors are involved in HIV-1 replication in macrophages. Previously, we have shown that genetic polymorphisms in phosphodiesterase 8a (PDE8A) are strongly associated with HIV-1 replication in these cells. Here we analyzed the mechanism and regulation of PDE8A in HIV-1 replication in macrophages. PDE8A mRNA expression strongly increases upon differentiation of monocytes into macrophages, which corresponds to the increased susceptibility of mature macrophages to HIV-1. In parallel, expression of microRNA miR-145-5p, predicted to target PDE8A mRNA, strongly decreased. The interaction of miR-145-5p with the 3' UTR of PDE8A mRNA could be experimentally validated, suggesting that indeed miR-145-5p can regulate PDE8A expression levels. Knockdown of PDE8A in macrophages resulted in a decrease in total HIV-1 replication and proviral DNA levels. These observations confirm that PDE8A regulates HIV-1 replication in macrophages and that this effect is mediated through early steps in the viral replication cycle. PDE8A is highly expressed in macrophages, and its expression is regulated by miR-145-5p. Our findings strongly suggest that PDE8A supports HIV-1 replication in macrophages and that this effect is mediated at the level of reverse transcription.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O