Merck
  • Metabolic profiling-based data-mining for an effective chemical combination to induce apoptosis of cancer cells.
Metabolic profiling-based data-mining for an effective chemical combination to induce apoptosis of cancer cells.
Scientific reports (2015-04-01)
Motofumi Kumazoe, Yoshinori Fujimura, Shiori Hidaka, Yoonhee Kim, Kanako Murayama, Mika Takai, Yuhui Huang, Shuya Yamashita, Motoki Murata, Daisuke Miura, Hiroyuki Wariishi, Mari Maeda-Yamamoto, Hirofumi Tachibana
ABSTRACT
Green tea extract (GTE) induces apoptosis of cancer cells without adversely affecting normal cells. Several clinical trials reported that GTE was well tolerated and had potential anti-cancer efficacy. Epigallocatechin-3-O-gallate (EGCG) is the primary compound responsible for the anti-cancer effect of GTE; however, the effect of EGCG alone is limited. To identify GTE compounds capable of potentiating EGCG bioactivity, we performed metabolic profiling of 43 green tea cultivar panels by liquid chromatography-mass spectrometry (LC-MS). Here, we revealed the polyphenol eriodictyol significantly potentiated apoptosis induction by EGCG in vitro and in a mouse tumour model by amplifying EGCG-induced activation of the 67-kDa laminin receptor (67LR)/protein kinase B/endothelial nitric oxide synthase/protein kinase C delta/acid sphingomyelinase signalling pathway. Our results show that metabolic profiling is an effective chemical-mining approach for identifying botanical drugs with therapeutic potential against multiple myeloma. Metabolic profiling-based data mining could be an efficient strategy for screening additional bioactive compounds and identifying effective chemical combinations.
Sigma-Aldrich
Naringenin, natural (US), 98%
Supelco
(±)-Naringenin, analytical standard
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Theobromine, ≥98.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
(±)-Naringenin, ≥95%
Alanine, European Pharmacopoeia (EP) Reference Standard
Supelco
Theobromine, analytical standard
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)
Sigma-Aldrich
Perchloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Hesperetin, ≥95%
Supelco
(−)-Epigallocatechin gallate, analytical standard
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Supelco
Hesperetin, analytical standard
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Epigallocatechin gallate, primary reference standard