Skip to Content
Merck
All Photos(1)

Key Documents

93894

Supelco

(−)-Epigallocatechin gallate

analytical standard

Synonym(s):

(−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-gallate, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxy-flavane-3-gallate, EGCG

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$373.00

$373.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$373.00

About This Item

Empirical Formula (Hill Notation):
C22H18O11
CAS Number:
Molecular Weight:
458.37
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$373.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@H](Oc2c1)c4cc(O)c(O)c(O)c4

InChI

1S/C22H18O11/c23-10-5-12(24)11-7-18(33-22(31)9-3-15(27)20(30)16(28)4-9)21(32-17(11)6-10)8-1-13(25)19(29)14(26)2-8/h1-6,18,21,23-30H,7H2/t18-,21-/m1/s1

InChI key

WMBWREPUVVBILR-WIYYLYMNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

(−)-Epigallocatechin gallate is a bioactive compound, which is an important constituent of green tea.[1] It may serve as a potent candidate to prevent cancer.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of tumor promoter-induced activator protein 1 activation and cell transformation by tea polyphenols,(-)-epigallocatechin gallate, and theaflavins
Dong Z, et al.
Cancer Research, 57, 4414-4419 (1997)
Mohsen Alizadeh et al.
Reports of biochemistry & molecular biology, 10(3), 455-461 (2022-01-05)
The available evidence has increasingly demonstrated that a combination of genetic and epigenetic factors, such as DNA methylation, could be considered as causing leukemia. Epigenetic changes and methylation of the suppressor of the cytokine signaling 1 promoter (SOCS1) CpG region
Epigallocatechin gallate has pleiotropic effects on transmembrane signaling by altering the embedding of transmembrane domains
Ye F, et al.
The Journal of Biological Chemistry, 292, 9858-9864 (2017)
Angélique Stalmach et al.
Molecular nutrition & food research, 53 Suppl 1, S44-S53 (2008-11-04)
Ten healthy human subjects consumed 500 mL of Choladi green tea, containing 648 mumol of flavan-3-ols after which plasma and urine were collected over a 24 h period and analysed by HPLC-MS. Plasma contained a total of ten metabolites, in
Hye-Kyung Na et al.
Molecular nutrition & food research, 50(2), 152-159 (2006-02-14)
Chemoprevention is an attempt to use either naturally occurring or synthetic substances or their mixtures to intervene in the progress of carcinogenesis. Recently, it has been shown that some edible phytochemicals alter gene expression, directly or indirectly, thereby regulating the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service