Merck
  • Quantitative analysis of zopiclone, N-desmethylzopiclone, zopiclone N-oxide and 2-amino-5-chloropyridine in urine using LC-MS-MS.
Quantitative analysis of zopiclone, N-desmethylzopiclone, zopiclone N-oxide and 2-amino-5-chloropyridine in urine using LC-MS-MS.
Journal of analytical toxicology (2014-05-03)
Gunnel H Nilsson, Fredrik C Kugelberg, Johan Ahlner, Robert Kronstrand
ABSTRACT
A simple liquid chromatography-tandem mass spectrometry method was validated to allow determination of zopiclone (ZOP), N-desmethylzopiclone (NDZOP), zopiclone N-oxide (ZOPNO) and 2-amino-5-chloropyridine (ACP) in urine at concentrations up to 3,000 ng/mL within 3.5 min. This method was used for quantitative analysis of the analytes in authentic urine samples obtained 10 h after oral administration of zopiclone (Imovane(®)) and in aliquots of the same urine samples after different storage conditions. In addition, pH of each studied urine sample was measured over time. The results showed that formation of ACP occurred at elevated pH and/or temperature by degradation of ZOP, NDZOP and ZOPNO. This method was also applied to samples obtained from two female victims of drug-facilitated assault. One sample had been exposed to long-term storage conditions at different temperatures and at pH >8.2, which resulted in high concentrations of ACP. The other sample, which was exposed to pH <6.5, showed no formation of ACP. ACP is formed both from ZOP and from its metabolites NDZOP and ZOPNO depending on the pH of the urine, time of storage and/or the temperature conditions. For correct interpretation in forensic cases, ZOP, its major metabolites and ACP should be analyzed. When ACP is identified in urine, the concentrations of ZOP, NDZOP and ZOPNO should be interpreted with great caution.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Methanol, analytical standard
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%