Merck
  • Supported liquid membrane-protected molecularly imprinted beads for micro-solid phase extraction of sulfonamides in environmental waters.
Supported liquid membrane-protected molecularly imprinted beads for micro-solid phase extraction of sulfonamides in environmental waters.
Journal of chromatography. A (2014-05-02)
M Díaz-Álvarez, F Barahona, E Turiel, A Martín-Esteban
ABSTRACT
In this work, molecularly imprinted polymer (MIP) beads have been prepared and evaluated for the development of a supported liquid membrane-protected micro-solid phase extraction method for the analysis of sulfonamides (SAs) in aqueous samples. The performance of MIP beads was firstly evaluated in cartridges by conventional solid-phase extraction for the simultaneous analysis of SAs. Afterward, beads were packed into a polypropylene hollow fiber protected by an organic solvent immobilized in the pores of the capillary wall. During the process, the analytes were extracted from the aqueous sample to the immobilized organic solvent and then selectively retained by the MIP beads located inside the capillary. The effect of various experimental parameters as sample pH, time and stirring-rate among others, were studied for the establishment of optimum rebinding conditions. Relative recoveries for all sulfonamides tested in river and reservoir water samples by the proposed method using 100mL water sample spiked with 50μg L-1 of each sulfonamide were within 70-120%, with a relative standard deviation (RSD) <10% (n=3). The detection limits (LODs) were within 0.2-3μgL(-1), depending upon the sulfonamide and the type of water used.
Product Description
Supelco
Sulfapyridine melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfadimethoxine, VETRANAL®, analytical standard
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Supelco
Sulfadimethoxine, 98.0-102.0%
Supelco
Sulfamonomethoxine, VETRANAL®, analytical standard
Sulfadimethoxine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfadimethoxine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfamonomethoxine
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Supelco
Sulfamethoxazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sulfamerazine, VETRANAL®, analytical standard
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Supelco
Aluminum oxide, for the determination of hydrocarbons
Supelco
Sulfapyridine, ≥99.0%
Supelco
Sulfamethoxazole
Sigma-Aldrich
Sulfamerazine, ReagentPlus®, ≥99.0%
Supelco
Sulfamethoxazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Sulfapyridine, VETRANAL®, analytical standard
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Sigma-Aldrich
Aluminum oxide, Brockmann I, activated, standard grade, free-flowing, Redi-Dri