Skip to Content
Merck
All Photos(1)

Key Documents

31737

Supelco

Sulfamethoxazole

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(5-methyl-3-isoxazolyl)benzenesulfonamide, N1-(5-Methylisoxazol-3-yl)sulfanilamide

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$80.40

$80.40


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$80.40

About This Item

Empirical Formula (Hill Notation):
C10H11N3O3S
CAS Number:
Molecular Weight:
253.28
Beilstein:
6732984
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$80.40


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cc1cc(NS(=O)(=O)c2ccc(N)cc2)no1

InChI

1S/C10H11N3O3S/c1-7-6-10(12-16-7)13-17(14,15)9-4-2-8(11)3-5-9/h2-6H,11H2,1H3,(H,12,13)

InChI key

JLKIGFTWXXRPMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfamethoxazole is a bacteriostatic antibiotic used in treating coccidiosis, diarrhea and gastroenteritis in humans and is also a known veterinary drug. Sulfamethoxazole in combination with trimethoprim is effective in treating pneumonia. Sulfamethoxazole acts by blocking the folic acid metabolism to produce synergistic antibacterial activity.[1][2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfamethoxazole may be used as a reference standard for the determination of the analyte in:
  • Bovine milk by high-performance liquid chromatography method.[2]
  • Pharmaceutical formulations and biological samples by capillary electrophoresis (CE) with end-column electrochemical detection (EC).[3]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatography method for the simultaneous determination of sulfamethoxazole and trimethoprim in bovine milk using an on-line clean-up column.
Pereira, AV and Cass, QB
Journal of Chromatography. B, Biomedical Sciences and Applications, 826(1-2), 139-146 (2005)
Sulfamethoxazole abatement by means of ozonation.
Dantas RF, et al.
Journal of Hazardous Materials, 150(3), 790-794 (2008)
Determination of sulfadiazine and sulfamethoxazole by capillary electrophoresis with end-column electrochemical detection.
You T, et al.
Analyst, 123(11), 2357-2360 (1998)
Ming Xie et al.
Water research, 47(13), 4567-4575 (2013-06-15)
The impact of humic acid fouling on the membrane transport of two pharmaceutically active compounds (PhACs) - namely carbamazepine and sulfamethoxazole - in forward osmosis (FO) was investigated. Deposition of humic acid onto the membrane surface was promoted by the
Karl J Schreiber et al.
BMC plant biology, 12, 226-226 (2012-11-28)
The sulfanilamide family comprises a clinically important group of antimicrobial compounds which also display bioactivity in plants. While there is evidence that sulfanilamides inhibit folate biosynthesis in both bacteria and plants, the complete network of plant responses to these compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service