Merck
  • Conjugated bisphenol A in maternal serum in relation to miscarriage risk.
Conjugated bisphenol A in maternal serum in relation to miscarriage risk.
Fertility and sterility (2014-04-22)
Ruth B Lathi, Cara A Liebert, Kathleen F Brookfield, Julia A Taylor, Frederick S vom Saal, Victor Y Fujimoto, Valerie L Baker
ABSTRACT
To examine the relationship between the maternal serum bisphenol A (BPA) concentration at the time of the missed menstrual cycle and miscarriage risk. Retrospective cohort of prospectively collected serum samples. Academic fertility center. Women presenting for early pregnancy monitoring with singleton pregnancies. Stored serum samples from 4 to 5 weeks' gestation analyzed for conjugated serum BPA concentrations. Live birth, miscarriage, and chromosome content of miscarriage. With the 115 women included in the study, there were 47 live births and 68 clinical miscarriages (46 aneuploid and 22 euploid). Median conjugated BPA concentrations were higher in the women who had miscarriages than in those who had live births (0.101 vs. 0.075 ng/mL). Women with the highest quartile of conjugated BPA had an increased relative risk of miscarriage (1.83; 95% CI, 1.14-2.96) compared with the women in the lowest quartile. We found a similar increase risk for both euploid and aneuploid miscarriages. Maternal conjugated BPA was associated with a higher risk of aneuploid and euploid miscarriage in this cohort. The impact of reducing individual exposure on future pregnancy outcomes deserves further study.
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
tert-Butyl methyl ether, analytical standard
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%