Merck
  • Synthesis of heterocyclic compounds through palladium-catalyzed C-H cyclization processes.
Synthesis of heterocyclic compounds through palladium-catalyzed C-H cyclization processes.
Chemical & pharmaceutical bulletin (2013-10-04)
Kiyofumi Inamoto
ABSTRACT
Herein, we describe our development of synthetic methods for heterocyclic compounds based on the palladium-catalyzed carbon-hydrogen bond (C-H) functionalization/intramolecular carbon-heteroatom (nitrogen or sulfur) bond formation process. By this C-H cyclization method, we efficiently prepared various N-heterocycles, including indazoles, indoles, and 2-quinolinones, as well as S-heterocycles such as benzothiazoles and benzo[b]thiophenes. Yields are typically good to high and good functional-group tolerance is observed for each process, thereby indicating that the method provides a novel, highly applicable synthetic route to the abovementioned biologically important heterocyclic frameworks. As an application of this approach, an auto-tandem-type, one-pot process involving the oxidative Heck reaction and subsequent C-H cyclization using cinnamamides and arylboronic acids as starting materials in the presence of a palladium catalyst was also developed for the rapid construction of the 2-quinolinone nucleus.
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Palladium, foil, not light tested, 25x25mm, thickness 0.0025mm, 99.95%
Palladium, microfoil, disks, 25mm, thinness 0.5μm, specific density 303.4μg/cm2, permanent mylar 3.5μm support, 99.99%
Palladium, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.95%
Palladium, rod, 50mm, diameter 2.0mm, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.02mm, as rolled, 99.95%
Palladium, microfoil, disks, 50mm, thinness 0.05μm, specific density 63.6μg/cm2, permanent mylar 3.5μm support, 99.99%
Palladium, foil, not light tested, 50x50mm, thickness 0.002mm, 99.95%
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.95%
Palladium, foil, 8mm disks, thickness 0.50mm, as rolled, 99.95%
Palladium, tube, 100mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.02mm, as rolled, 99.95%
Palladium, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95%
Palladium, wire reel, 10m, diameter 0.025mm, as drawn, 99.9%
Palladium, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.75%
Palladium, wire reel, 50m, diameter 0.125mm, hard, 99.95%