Skip to Content
Merck
All Photos(2)

Key Documents

GF52273852

Palladium

wire reel, 10m, diameter 0.025mm, as drawn, 99.9%

Synonym(s):

Palladium, PD005113

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$721.00

$721.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$721.00

About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
MDL number:
UNSPSC Code:
12141733
PubChem Substance ID:
NACRES:
NA.23

$721.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 522-738-52

resistivity

9.96 μΩ-cm, 20°C

L × diam.

10 m × 0.025 mm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Update 1 of: Synthesis and functionalization of indoles through palladium-catalyzed reactions.
Sandro Cacchi et al.
Chemical reviews, 111(5), PR215-PR283 (2011-05-12)
Amanda J Hickman et al.
Nature, 484(7393), 177-185 (2012-04-14)
Copper and palladium catalysts are critically important in numerous commercial chemical processes. Improvements in the activity, selectivity and scope of these catalysts could drastically reduce the environmental impact, and increase the sustainability, of chemical reactions. One rapidly developing strategy for
Synthesis of heterocycles via palladium-catalyzed carbonylations.
Xiao-Feng Wu et al.
Chemical reviews, 113(1), 1-35 (2012-10-09)
Anton V Dubrovskiy et al.
Combinatorial chemistry & high throughput screening, 15(6), 451-472 (2012-01-26)
The iodocyclization of functionally-substituted alkynes provides an excellent way to prepare a wide range of iodoheterocycles, which can then be readily elaborated through palladium-catalyzed Suzuki-Miyaura, Sonogashira, Heck, Hartwig-Buchwald, and carbonylation processes into libraries of medicinally relevant heterocycles. The synthesis of
Jana Doháňošová et al.
Molecules (Basel, Switzerland), 18(6), 6173-6192 (2013-05-28)
The palladium (II)-catalysed reactions of alkenols and aminoalkenols such as oxycarbonylations or bicyclisations are powerful methods for the construction of oxygen and nitrogen-containing heterocyclic compounds. This review highlights recent progress in the development of the asymmetric palladium(II)-catalysed Wacker-type cyclisations of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service