Merck
Recombinant collagen and gelatin for drug delivery.
Advanced drug delivery reviews (2003-11-19)
David Olsen, Chunlin Yang, Michael Bodo, Robert Chang, Scott Leigh, Julio Baez, David Carmichael, Maritta Perälä, Eija-Riitta Hämäläinen, Marko Jarvinen, James Polarek
ABSTRACT
The tools of recombinant protein expression are now being used to provide recombinant sources of both collagen and gelatin. The primary focus of this review is to discuss alternatives to bovine collagen for biomedical applications. Several recombinant systems have been developed for production of human sequence collagens. Mammalian and insect cells were initially used, but were thought to be too costly for commercial production. Yeast have been engineered to express high levels of type I homotrimer and heterotrimer and type II and type III collagen. Co-expression of collagen genes and cDNAs encoding the subunits of prolyl hydroxylase has lead to the synthesis of completely hydroxylated, thermostable collagens. Human types I and III collagen homotrimers have been expressed in transgenic tobacco plants, while transgenic mice have been engineered to produce full-length type I procollagen homotrimer as well as a alpha2 (I) homotrimeric mini-collagen. Most recently, a transgenic silkworm system was used to produce a fusion protein containing a collagenous sequence. Each of these transgenic systems holds great promise for the cost-effective large-scale production of recombinant human collagens. As seen in other recombinant expression systems, transgenic silkworms, tobacco, and mice lack sufficient endogenous prolyl hydroxylase activity to produce fully hydroxylated collagen. In mice and tobacco, this was overcome by over-expression of prolyl hydroxylase, analogous to what has been done in yeast and insect cell culture. In addition to recombinant alternatives to bovine collagen, other sources such as fish and sponge collagen are discussed briefly. Recombinant gelatin has been expressed in Pichia pastoris and Hansenula polymorpha in both non-hydroxylated and hydroxylated forms. Pichia was shown to be a highly productive system for gelatin production. The recombinant gelatins produced in yeast are of defined molecular weight and physio-chemical properties and represent a new biomaterial not previously available from animal sources. Genetic engineering has made great progress in the areas of recombinant collagen and gelatin expression, and there are now several alternatives to bovine material that offer an enhanced safety profile, greater reproducibility and quality, and the ability of these materials to be tailored to enhance product performance.
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength