Skip to Content
Merck
All Photos(1)

Key Documents

48724

Millipore

Gelatin from porcine skin

suitable for microbiology, high gel strength

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$115.00

$115.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$115.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

$115.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

biological source

Porcine

Quality Level

sterility

non-sterile

form

powder

quality

high gel strength

shelf life

limited shelf life, expiry date on the label

ign. residue

≤2%

loss

≤15% loss on drying

transmittance

450 nm, ≥85%
620 nm, ≥95%

pH

4.0-6.0 (25 °C, 67 mg/mL in H2O)
5.20-5.60

viscosity

5.10-5.80 mPa.s

gel strength

245-275 g Bloom (67 mg/ml water)
280-320 g BloomAOAC

solubility

H2O: 67 mg/mL at 50 °C, slightly hazy, faintly yellow
H2O: 67 mg/mL at 60 °C

anion traces

chloride (Cl-): ≤2000 mg/kg

cation traces

Ca: ≤2000 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤10 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤500 mg/kg
Mg: ≤500 mg/kg
Mn: ≤5 mg/kg
Na: ≤1200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

application(s)

microbiology

Looking for similar products? Visit Product Comparison Guide

Application

This product is recommended for use as a cell culture substratum at 1-5 μg/cm2 or 0.5-50 μg/mL. The optimal concentration does depend on cell type as well as the application and research objectives.

Gelatin has been used in many applications. It has use in coating cell culture to improve attachment of cells, being added to PCR to stabilize Taq DNA, as a blocking reagent in Western blotting, ELISA, and immunochemistry, and as a component of media for species differentiation in bacteriology. As a biocompatible polymer, it has used as a delivery vehicle for release of active biomolecules and in generation of scaffolds for tissue engineering applications. In the pharmaceutical industry, geltan can be used as a suspending and encapsulating agent, among other applications.

Components

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue.

Caution

Dry gelatin, when stored in airtight containers at room temperature, will remain unchanged for many years. When heated at 100°C in the presence of air, it swells becomes soft and disintegrates to a carbonaceous mass with evolution of pyridine bases and ammonia.

Preparation Note

This product is derived from porcine skin. Gelatin is soluble in hot than in cold water. It is practically insoluble in most organic solvents such as alcohol, chloroform, carbon disulfide, carbon tetrachloride, ether, benzene, acetone, and oils. The Bloom number, determined by the Bloom gelometer, is an indication of the strength of a gel formed from a solution of the known concentration. The Bloom number is proportional to the average molecular mass. Bloom numbers of porcine skin Gelatin vary from 90 to 300 g. This product has a gel strength of 250. Manufactured by Gelita AG

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Busk Jr., G.C.
Food Technology, 38, 59-59 (1984)
Kozlov, P.V., and Burdygina, G.I.
Polymer, 24, 651-651 (1983)
Nur Cebi et al.
Food chemistry, 277, 373-381 (2018-12-07)
Gelatin is widely used in gummy candies because of its unique functional properties. Generally, porcine and bovine gelatins are used in the food industry. FTIR-ATR combined with chemometrics analysis such as hierarchical cluster analysis (HCA) (OPUS Version 7.2 software), principal
Leslie Crews et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(16), 4250-4260 (2008-04-18)
Altered expression and mutations in alpha-synuclein (alpha-syn) have been linked to Parkinson's disease (PD) and related disorders. The neurological alterations in PD patients have been associated with degeneration of dopaminergic cells and other neuronal populations. Moreover, recent studies in murine
Louise Clemmesen et al.
Ugeskrift for laeger, 175(9), 576-578 (2013-04-24)
This article describes the production of a low-cost training phantom for ultrasound guided invasive procedures of peripheral and central veins and presents a video of the process. The phantom can be adapted for use with other ultrasound techniques. It is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service