Merck
  • The effect of increased concentrations of homocysteine on the concentration of (E)-4-hydroxy-2-nonenal in the plasma and cerebrospinal fluid of patients with Alzheimer's disease.
The effect of increased concentrations of homocysteine on the concentration of (E)-4-hydroxy-2-nonenal in the plasma and cerebrospinal fluid of patients with Alzheimer's disease.
Neurobiology of aging (2002-04-18)
M L Selley, D R Close, S E Stern
ABSTRACT
There is evidence that increased blood concentrations of homocysteine may be a risk factor for Alzheimer's disease. (E)-4-hydroxy-2-nonenal (HNE) is a neurotoxic product of lipid peroxidation that is increased in the ventricular fluid and brains of patients with Alzheimer's disease. We measured the concentrations of homocysteine, HNE, vitamin B(12) and folate in the plasma of 27 patients with Alzheimer's disease and 25 control subjects. There was a statistically significant increase in the plasma concentration of homocysteine (P < 0.001) and HNE (P < 0.001) in the Alzheimer's disease patients compared to the control group. There was a significant decrease in the plasma concentration of vitamin B(12) (P < 0.001) and folate (P = 0.002) in the Alzheimer's group compared to the controls. There was a significant positive correlation between the plasma concentrations of homocysteine and HNE in the patients with Alzheimer's disease (r = 0.661, P < 0.001). A significant negative correlation was found between the plasma concentration of homocysteine and the plasma concentrations of vitamin B(12) (r = -0.605, P = 0.0006) and folate (r = 0.586, P = 0.001). We also measured the concentrations of homocysteine, HNE, vitamin B(12) and folate in the cerebrospinal fluid (CSF) of 8 patients with Alzheimer's disease compared to 6 control subjects. The concentrations of homocysteine (P = 0.032) and HNE (P = 0.001) were significantly higher in the CSF of Alzheimer's patients than in the control subjects. There were significant positive correlations between the CSF concentrations of homocysteine and HNE (r = 0.924, P = 0.001). There was also a significant positive correlation between the plasma concentration of homocysteine and the CSF concentrations of homocysteine (r = 0.850, P = 0.007) and HNE (r = 0.092, P = 0.002). These results demonstrate that there is a relationship between increased homocysteine concentrations and increased HNE concentrations in Alzheimer's disease.
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Millipore
myo-Inositol, ≥99.0%, suitable for microbiology
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)