Skip to Content
Merck
All Photos(1)

Documents

348791

Sigma-Aldrich

Titanium

foil, thickness 0.127 mm, 99.7% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
EC Number:
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.7% trace metals basis

form

foil

autoignition temp.

860 °F

resistivity

42.0 μΩ-cm, 20°C

thickness

0.127 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

Application

Titanium based foils can be used in the anodization process for the fabrication of titanium nanotubes (TNTs) which can further be used in solar cells and other sensor based applications.

Quantity

150 × 150 mm (approximately 13 g)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fabrication of high aspect ratio and open-ended TiO2 nanotube photocatalytic arrays through electrochemical anodization
Wong RJ, et al.
AIChE Journal, 62(2), 415-420 (2016)
A new approach to electropolishing of pure Ti foil in acidic solution at room temperature for the formation of ordered and long TiO2 nanotube arrays
Asgari V, et al.
Corrosion Science, 136(6), 38-46 (2018)
N-doped TiO 2 nanotubes coated with a thin TaO x N y layer for photoelectrochemical water splitting: dual bulk and surface modification of photoanodes
Kim H, et al.
Energy & Environmental Science, 8(1), 247-257 (2015)
Rapid TiO 2 Nanotubes Formation in Aged Electrolyte and Their Application as Photocatalysts for Cr (VI) Reduction Under Visible Light
Zulkifli MA, et al.
Nanotechnology, IEEE Transactions, 17(6), 1106-1110 (2018)
Jinho Shin et al.
Journal of nanoscience and nanotechnology, 13(8), 5807-5810 (2013-07-26)
In this study, hydroxyapatite (HA) was coated on anodized titanium (Ti) surfaces through radio frequency magnetron sputtering in order to improve biological response of the titanium surface. All the samples were blasted with resorbable blasting media (RBM). RBM-blasted Ti surface

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service