Merck
  • Batch and column study: sorption of perfluorinated surfactants from water and cosolvent systems by Amberlite XAD resins.
Batch and column study: sorption of perfluorinated surfactants from water and cosolvent systems by Amberlite XAD resins.
Journal of colloid and interface science (2011-12-14)
Feng Xiao, Kerry Jade Davidsavor, Sangyoo Park, Michio Nakayama, Brian Ray Phillips
ABSTRACT
Amberlite XAD resins have been employed to a great extent as the sorbent for removing or concentrating organic compounds from different matrices. We present for the first time a systematic study on the sorption of perfluorochemical (PFC) surfactants, an emerging class of environmental contaminants, by XAD-7HP (moderately polar) and XAD-2 (nonpolar). The results show that XAD-7HP can strongly sorb PFCs at circumneutral pH; the isotherm-determined linear sorption coefficient can reach 10(6)L/kg. On the other hand, the sorption coefficient for XAD-2 was two orders of magnitude lower than that for XAD-7HP. PFC sorption on XAD-7HP increased with an increase of the perfluorocarbon chain length of PFC and a decrease of the solution pH, indicating the importance of hydrophobic and electrostatic effects. The sorption coefficient for XAD-7HP reduced markedly with increasing fraction of the organic cosolvent (methanol) in the water-cosolvent mixture; however, the trend could not be predicted by a log-linear cosolvency model. Furthermore, the statistical analysis of column test results showed that after regeneration XAD-7HP can be used at least eight times without significant loss of performance. Finally, the experimental results imply that XAD-7HP sorption of shorter-chained PFCs (≤5 perfluorinated carbons) from water can be thermodynamically favorable.
Product Description
Supelco
Amberlite XAD®-2, pkg of 100 g
Supelco
Amberlite XAD®-2, pkg of 25 kg
Supelco
Amberlite XAD®-2, pkg of 10 kg
Supelco
Amberlite XAD®-2, pkg of 500 g
Supelco
Amberlite XAD®-2, pkg of 5 kg