Skip to Content
Merck
All Photos(1)

Documents

52672-U

Supelco

Amberlite XAD®-2 polymeric adsorbent

20-60 mesh, pkg of 10 kg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000
NACRES:
SB.52

product name

Amberlite XAD®-2, pkg of 10 kg

form

solid

packaging

pkg of 10 kg

availability

not available in (Currently experiencing backorder delays due to raw material supply disruption)

technique(s)

HPLC: suitable
LPLC: suitable

surface area

~300 m2/g

matrix

styrene-divinylbenzene (macroreticular)

matrix active group

polymer

particle size

20-60 mesh

pore size

~0.65 mL/g pore volume
90 Å mean pore size

density

1.02 g/mL at 25 °C (true wet)(lit.)
1.08 g/mL at 25 °C (skeletal)(lit.)

separation technique

reversed phase

InChI

1S/C24H27NO5/c1-24(2,3)30-22(27)13-12-16(14-26)25-23(28)29-15-21-19-10-6-4-8-17(19)18-9-5-7-11-20(18)21/h4-11,14,16,21H,12-13,15H2,1-3H3,(H,25,28)/t16-/m0/s1

InChI key

BCIPGSZQUDLGSY-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite XAD-2, a hydrophobic copolymer of styrene-divinylbenzene resin which requires longer purification process and is used as a sorbent. Amberlite XAD-2 resin depending upon the aromaticity and size of the hydrophobic groups sorbs organic compounds through low-energy physical interactions.
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
Specifications:

Application

Amberlite XAD®-2 was used:
  • as adsorbent in the extraction of organochlorine compounds using Soxhlat extraction technique
  • as adsorbent in the extraction of honey samples using column chromatography for HPLC analysis
  • in adsorption chromatography
Polyaromatic adsorbent resin for hydrophobic compounds up to MW 20,000: phenols, organic removal, surfactants, aroma compounds, antibiotic recovery. May be used as support for catalyst or metals removal. For cleaned US EPA versions, see Supelpak-2 and Supelpak-2B.
     

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trapping efficiency of polyurethane foam and Amberlite XAD-2 for various organochlorine compounds
Nerin, C., et al.
Fresenius Journal of Analytical Chemistry, 352, 609-611 (1995)
I Martos et al.
Journal of agricultural and food chemistry, 48(5), 1498-1502 (2000-05-23)
European Eucalyptus honeys showed a common and characteristic HPLC profile in which the flavonoids myricetin (3,5,7,3',4', 5'-hexahydroxyflavone), tricetin (5,7,3',4',5'-pentahydroxyflavone), quercetin (3,5,7,3',4'-pentahydroxyflavone), luteolin (5,7,3', 4'-tetrahydroxyflavone), and kaempferol (3,5,7, 4'-tetrahydroxyflavone) were identified. Their contents, and relative amounts, in the analyzed honey samples
M Tamura et al.
The Journal of biological chemistry, 260(17), 9672-9677 (1985-08-15)
Na+,K+-ATPase inhibitors have been found to exist in acutely saline-infused hog plasma, which also inhibit the specific binding of ouabain to Na+,K+-ATPase and the binding of digoxin to specific anti-digoxin antibody. Two of these inhibitors were purified by a combination
Sorption of sodium dodecyl sulfate on amberlite XAD-2 resin.
HIRAIDE, Masataka, Toshihiko SHIMA, and Hiroshi KAWAGUCHI.
Analytical Sciences, 10, 505-507 (1994)
Application of Amberlite XAD resins to the isolation of humic substances from sea water.
Grzybowski, W.
Oceanologia, 33, 105-109 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service