Merck
  • Hydrocarbon biodegradation and dynamic laser speckle for detecting chemotactic responses at low bacterial concentration.
Hydrocarbon biodegradation and dynamic laser speckle for detecting chemotactic responses at low bacterial concentration.
Journal of environmental sciences (China) (2013-08-08)
Melina Nisenbaum, Gonzalo Hernán Sendra, Gastón Alfredo Cerdá Gilbert, Marcelo Scagliola, Jorge Froilán González, Silvia Elena Murialdo
ABSTRACT
We report on the biodegradation of pure hydrocarbons and chemotaxis towards these compounds by an isolated chlorophenol degrader, Pseudomonas strain H. The biochemical and phylogenetic analysis of the 16S rDNA sequence identified Pseudomonas strain H as having 99.56% similarity with P. aeruginosa PA01. This strain was able to degrade n-hexadecane, 1-undecene, 1-nonene, 1-decene, 1-dodecene and kerosene. It grew in the presence of 1-octene, while this hydrocarbons is toxic to other hydrocarbons degraders. Pseudomonas strain H was also chemotactic towards n-hexadecane, kerosene, 1-undecene and 1-dodecene. These results show that this Pseudomonas strain H is an attractive candidate for hydrocarbon-containing wastewater bioremediation in controlled environments. Since the classical standard techniques for detecting chemotaxis are not efficient at low bacterial concentrations, we demonstrate the use of the dynamic speckle laser method, which is simple and inexpensive, to confirm bacterial chemotaxis at low cell concentrations (less than 10(5) colony-forming unit per millilitre (CFU/mL)) when hydrocarbons are the attractants.
Product Description
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Supelco
Activated charcoal, puriss. p.a., powder
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, acid-washed with phosphoric and sulfuric acids
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
1-Dodecene, ≥99.0% (GC)
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder