Skip to Content
Merck
All Photos(2)

Key Documents

31616

Supelco

Activated Charcoal Carbon Adsorbent

greener alternative

puriss. p.a., powder

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$87.60
750 G
$227.00

$87.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$87.60
750 G
$227.00

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.54

$87.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Activated charcoal, puriss. p.a., powder

vapor pressure

<0.1 mmHg ( 20 °C)

Quality Level

description

methylene blue-adsorption 12 mL

grade

puriss. p.a.

form

powder

autoignition temp.

842 °F

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

1375 μΩ-cm, 20°C (graphite)

technique(s)

LPLC: suitable
tissue culture: suitable

impurities

≤0.003% heavy metals (as Pb)
≤0.5% in water soluble matter
≤1% soluble in acid

ign. residue

≤2%

loss

≤8% loss on drying, 120 °C

matrix

Carbon

matrix active group

carbon

pH

6-9

mp

3550 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Cu: ≤25 mg/kg
Fe: ≤300 mg/kg
Pb: ≤10 mg/kg
Zn: ≤10 mg/kg

greener alternative category

separation technique

reversed phase

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Activated charcoal is an FDA approved adsorbent and generally finds application in being used during the treatment of swallowed poison.[1]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated charcoal can be used for the following:
  • to study its effect on tissue cultures of Daucus carota, Happlopapus gracilis and Allum cepa var. proliferum, to check the adsorption of metabolites inhibiting morphognesis.[2]
  • It may be used as the primary gastrointestinal decontamination procedure after acute drug overdose.[3]
  • It may also be used for the adsorption of dyes namely malachite green, bromophenol blue, alizarine red-S, methylene blue, eriochrome black-T, phenol red, methyl blue, and methyl violet from aqueous media.[4]
  • Oral activated charcoal may be also used for adsorption of theophylline, hence can be effectively used in patients with theophylline poisoning.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activated Carbon Applications in the Food and Pharmaceutical Industries.
Roy GM.
Technology & Engineering, 137-138 (1994)
The effect of activated charcoal on tissue cultures: adsorption of metabolites inhibiting morphogenesis.
Gunnar F
Physiologia Plantarum, 43 (2), 104-106 (1978)
Muhammad J Iqbal et al.
Journal of hazardous materials, 139(1), 57-66 (2006-07-20)
Adsorption of industrially important dyes namely bromophenol blue, alizarine red-S, methyl blue, methylene blue, eriochrome black-T, malachite green, phenol red and methyl violet from aqueous media on activated charcoal has been investigated. The effect of shaking time, pH and temperature
M Tenenbein et al.
Annals of emergency medicine, 16(8), 838-841 (1987-08-01)
The efficacy of ipecac-induced emesis, large-bore orogastric lavage, and activated charcoal as gastrointestinal decontamination procedures after acute drug overdose is unknown. Using an ampicillin overdose model, these three procedures were compared with one another and to a control ingestion in
W G Berlinger et al.
Clinical pharmacology and therapeutics, 33(3), 351-354 (1983-03-01)
A randomized crossover trial of the effect of oral activated charcoal on the kinetics of intravenous theophylline was carried out in six normal male subjects. After intravenous aminophylline (6 mg/kg), subjects received water or water with activated charcoal (140 gm)

Questions

1–2 of 2 Questions  
  1. Is this activate charcoal phosphate-free? It's called for in soil phosphorous measurements

    1 answer
    1. There is no specific phosphate test for this product. This means while the activated charcoal is commonly used in applications such as soil phosphorus measurements, there is no guarantee that this product is phosphate-free.

      Helpful?

  2. What is the surface area of this activated carbon?

    1 answer
    1. The total surface area for this product is 1050-1150 m²/g.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service