The total surface area for this product is 875 m²/g.
22874
Activated Charcoal Norit®
Norit® PK 1-3, from peat, steam activated, granular
Synonym(s):
Charcoal activated Norit®
Select a Size
About This Item
Recommended Products
biological source
peat
form
granular
quality
steam activated
manufacturer/tradename
Norit® PK 1-3
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
particle size
1-3 mm
greener alternative category
, Enabling
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the surface area of this activated carbon?
1 answer-
Helpful?
-
-
I would like to know the surface area and the pore size of this product?
1 answer-
Unfortunately, the surface area and pore size of Product 22874 are not tested as per the associated Certificate of Analysis. There is no lot specific value available. The reference value is 875 m2/g for surface area.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service