Merck
  • Influence of fuel mass load, oxygen supply and burning rate on emission factor and size distribution of carbonaceous particulate matter from indoor corn straw burning.
Influence of fuel mass load, oxygen supply and burning rate on emission factor and size distribution of carbonaceous particulate matter from indoor corn straw burning.
Journal of environmental sciences (China) (2013-08-08)
Guofeng Shen, Miao Xue, Siye Wei, Yuanchen Chen, Bin Wang, Rong Wang, Huizhong Shen, Wei Li, Yanyan Zhang, Ye Huang, Han Chen, Wen Wei, Qiuyue Zhao, Bin Li, Haisuo Wu, Shu Tao
ABSTRACT
The uncertainty in emission estimation is strongly associated with the variation in emission factor (EF), which could be influenced by a variety of factors such as fuel properties, stove type, fire management and even methods used in measurements. The impacts of these factors are complicated and often interact with each other. Controlled burning experiments were conducted to investigate the influences of fuel mass load, air supply and burning rate on the emissions and size distributions of carbonaceous particulate matter (PM) from indoor corn straw burning in a cooking stove. The results showed that the EFs of PM (EF(PM)), organic carbon (EFoc) and elemental carbon (EF(EC)) were independent of the fuel mass load. The differences among them under different burning rates or air supply amounts were also found to be insignificant (p > 0.05) in the tested circumstances. PM from the indoor corn straw burning was dominated by fine PM with diameter less than 2.1 microm, contributing 86.4% +/- 3.9% of the total. The size distribution of PM was influenced by the burning rate and air supply conditions. On average, EF(PM), EF(OC) and EF(EC) for corn straw burned in a residential cooking stove were (3.84 +/- 1.02), (0.846 +/- 0.895) and (0.391 +/- 0.350) g/kg, respectively. EF(PM), EF(OC) and EF(EC) were found to be positively correlated with each other (p < 0.05), but they were not significantly correlated with the EF of co-emitted CO, suggesting that special attention should be paid to the use of CO as a surrogate for other incomplete combustion pollutants.
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Supelco
Activated charcoal, powder
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, acid-washed with phosphoric and sulfuric acids
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod