Merck
  • Dextran-functionalized magnetic fluid mediating magnetohyperthermia combined with preventive antioxidant pequi-oil supplementation: potential use against cancer.
Dextran-functionalized magnetic fluid mediating magnetohyperthermia combined with preventive antioxidant pequi-oil supplementation: potential use against cancer.
Journal of biomedical nanotechnology (2013-08-06)
Ana Luisa Miranda-Vilela, Raphael Cândido Apolinário Peixoto, Joāo Paulo Figueiró Longo, Débora de Oliveira Silva e Cintra, Flávia Arruda Portilho, Kely Lopes Caiado Miranda, Patrícia Pommé Confessori Sartoratto, Sônia Nair Báo, Ricardo Bentes de Azevedo, Zulmira Guerrero Marques Lacava
ABSTRACT
This work aimed to test a dextran-functionalized magnetic fluid (DexMF) sample in mediating magnetohyperthermia to treat an advanced clinical Ehrlich-solid-tumor, to verify the effects of oral antioxidant administration of pequi-oil on this treatment and to investigate the potential of these treatments for future use as an adjuvant in cancer therapy. Animals received the treatments: (a) filtered water (control); (b) tumor implantation and no treatment (tumor group); (c) tumor implantation followed by intratumoral injection of DexMF and alternating current magnetic field exposure (MHT group) for three consecutive days; (d) oral pequi-oil supplementation followed by tumor implantation and the same treatment as group MHT (PMHT group). Analyses took place 1 and 2 weeks after tumor implantation. Both treatments were effective in increasing the tumor necrosis process and controlling tumor growth, besides keeping lymphocyte-dependent immunity. Although the MHT treatment was more efficient after the first week in reducing DNA damage to blood peripheral leucocytes, PMHT therapy appeared to be more effective with the advance of the carcinogenesis process after the second week. Our findings evidence the potential use of DexMF mediating magnetohyperthermia in cancer treatment and also suggest that the preventive pequi oil administration could increase the efficiency of this process.
Product Description
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 670,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 1,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 12,000
Supelco
Dextran, analytical standard, Set Mp 1,000-400,000
Supelco
Dextran, analytical standard, 5,000
Supelco
Dextran, analytical standard, 25,000
Supelco
Dextran, analytical standard, 410,000
Supelco
Dextran, analytical standard, 150,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Supelco
Dextran, analytical standard, 670,000
Supelco
Dextran, analytical standard, 80,000
Supelco
Dextran, analytical standard, 50,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Supelco
Dextran, analytical standard, 1,000
Supelco
Dextran, analytical standard, 270,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000