Merck
  • Microwave-enhanced Fenton-like system, Cu(II)/H2O2, for olive mill wastewater treatment.
Microwave-enhanced Fenton-like system, Cu(II)/H2O2, for olive mill wastewater treatment.
Environmental technology (2013-07-11)
Hamida Iboukhoulef, Abdeltif Amrane, Hocine Kadi
ABSTRACT
Olive mill wastewater (OMW) is a substantial pollutant not only because of its high organic matter content and recalcitrant compounds such as polyphenols, but also due to its high chemical oxygen demand and biological oxygen demand. Its degradation by means of hydroxyl radicals generated from microwave application in the presence of hydrogen peroxide and Cu(II) has been investigated. The optimal experimental conditions were found to be a microwave intensity of 680 W, the presence of Cu(II) and relatively concentrated hydrogen peroxide (12 mol L(-1)). In these conditions, discolouration of OMW reached 98.0 +/- 0.2% and phenolic compounds decreased to 81.8 +/- 0.2% of their initial concentration after 12 min microwave time. Fourier transform infrared analysis confirmed that microwave degradation of phenols by means of the Fenton-like system Cu(II)/H2O2 could be an efficient solution for the treatment of olive mill wastewater.
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Continuous cast copper (O), BCR®, certified reference material, rod
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Copper (S, P), BCR®, certified reference material, chips
Copper, IRMM®, certified reference material, 0.5 mm wire
Copper, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%