IRMM522A
Copper
IRMM®, certified reference material, 0.1 mm foil
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
Agency
IRMM®
manufacturer/tradename
JRC
resistivity
1.673 μΩ-cm, 20°C
bp
2567 °C (lit.)
mp
1083.4 °C (lit.)
density
8.94 g/mL at 25 °C (lit.)
format
matrix material
SMILES string
[Cu]
InChI
1S/Cu
InChI key
RYGMFSIKBFXOCR-UHFFFAOYSA-N
Application
Copper may be used as a standard reference material in studying the thermal expansion coefficient of different available grades of copper like high purity copper, tough pitch copper and oxygen-free high-conductivity copper using absolute laser interferometric dilatometer.
Analysis Note
For more information please see:
IRMM522A
IRMM522A
Legal Information
IRMM is a registered trademark of European Commission
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Measurements of linear thermal expansion coefficients of copper SRM 736 and some commercially available coppers in the temperature range 20?300 K by means of an absolute interferometric dilatometer
Cryogenics, 37(5), 251-254 (1997)
Chimia, 67(6), 383-387 (2013-08-16)
The node-and-spacer approach is widely employed in crystal engineering to construct coordination polymers. It consists of self-assembly processes involving mononuclear cationic species and exo-dentate ligands. We enlarged this strategy using preformed homo- and heterometallic complexes as nodes. The presence of
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 96(2), 150-156 (2013-08-14)
Wilson's disease (WD) is an inherited disorder of copper metabolism, leading to hepatic and neuropsychiatric manifestations. The long-term outcome of patients with Wilson's disease presenting with acute liver failure (ALF) versus those with acute-on-chronic liver failure (ACLF) has not been
Journal of environmental sciences (China), 25(4), 652-658 (2013-08-09)
The removal efficiency of copper (Cu(II)) from an actual acidic electroplating effluent by biochars generated from canola, rice, soybean and peanut straws was investigated. The biochars simultaneously removed Cu(II) from the effluent, mainly through the mechanisms of adsorption and precipitation
Journal of environmental sciences (China), 25(4), 808-814 (2013-08-09)
Metal (Cu, Co, or Zn) loaded ZSM-5 and Y zeolite adsorbents were prepared for the adsorption of hydrogen cyanide (HCN) toxic gas. The results showed that the HCN breakthrough capacity was enhanced significantly when zeolites were loaded with Cu. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service