Skip to Content
Merck
All Photos(1)

Documents

BCR058

Continuous cast copper (O)

BCR®, certified reference material, rod

Synonym(s):

Copper

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
CAS Number:
Molecular Weight:
63.55
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

form

rod

manufacturer/tradename

JRC

resistivity

1.673 μΩ-cm, 20°C

bp

2567 °C (lit.)

mp

1083.4 °C (lit.)

density

8.94 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

[Cu]

InChI

1S/Cu

InChI key

RYGMFSIKBFXOCR-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR058

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marius Andruh
Chimia, 67(6), 383-387 (2013-08-16)
The node-and-spacer approach is widely employed in crystal engineering to construct coordination polymers. It consists of self-assembly processes involving mononuclear cationic species and exo-dentate ligands. We enlarged this strategy using preformed homo- and heterometallic complexes as nodes. The presence of
Ping Ning et al.
Journal of environmental sciences (China), 25(4), 808-814 (2013-08-09)
Metal (Cu, Co, or Zn) loaded ZSM-5 and Y zeolite adsorbents were prepared for the adsorption of hydrogen cyanide (HCN) toxic gas. The results showed that the HCN breakthrough capacity was enhanced significantly when zeolites were loaded with Cu. The
Ren Zhang et al.
Chimia, 67(6), 393-396 (2013-08-16)
Three new R-isophthalic acid-based (R = H or Br) coordination polymers have been designed and synthesized. By changing the N-containing ligand in the system, we are able to tune the dimensionality of coordination polymers from one-dimension (1D) to two-dimensions (2D)
A Turgut et al.
European review for medical and pharmacological sciences, 17(11), 1472-1478 (2013-06-19)
To compare patients with advanced stage endometriosis with control patients without endometriosis with respect to serum Copper (Cu) and Ceruloplasmin (Cp) levels and oxidative stress markers in order to evaluate the importance of these parameters in the pathogenesis of endometriosis.
John Mondal et al.
Journal of nanoscience and nanotechnology, 13(7), 4883-4895 (2013-08-02)
Furfural-imine functionalized mesoporous organosilica material has been synthesized by post-synthesis surface functionalization of 2D-hexagnoal mesoporous SBA-15 with organosilane precursor 3-aminopropyltriethoxy-silane (APTES) followed by Schiff-base condensation with furfural. On the other hand, furfural-imine functionalized MCM-41 has been synthesized by Schiff-base condensation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service