Merck
  • [To a question on the impact of nanoparticles of metals present in the aquatic environment, on bacteria and continuous cell lines HEp-2 and BGM].
[To a question on the impact of nanoparticles of metals present in the aquatic environment, on bacteria and continuous cell lines HEp-2 and BGM].
Gigiena i sanitariia (2013-06-29)
A V Zagainova, T Z Artemova, R A Dmitrieva, E K Gipp, L V Ivanova, Yu G Talaeva, N N Butorina, T N Maksimkina, O V Savostikova, O V Ushakova, M A Vodianova, F I Ingel, V V Yurchenko, E K Krivtsova
ABSTRACT
Promising application of nanoparticles and nanomaterials is the creation of sanitary hygienic means of new generation used for disinfection of water and indoor surfaces of mass use, furniture, sanitary technical equipment by virtue of modifying traditional materials to bring them effective biocidal properties, and for the development of methods in vitro for assessment their toxicity In this paper the possibility of the use various forms of silver, copper and aluminum as disinfectant for bacterial test organisms in the aquatic environment and assess their toxicity on biological models of continuous culture of BGM cells (a stable line of African green monkey kidney cells) and HEp-2 (Human epithelial type 2 (HEp-2) cells, derived from a human laryngeal carcinoma) is considered.
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Copper, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%